EMAIL THIS PAGE TO A FRIEND
International journal of environmental health research
Role of access to parks and markets with anthropometric measurements, biological markers, and a healthy lifestyle.
PMID 25236622
Abstract
This study examined the association between access to urban green spaces and markets with anthropometric measurements, biological markers, sociodemographic, and healthy lifestyle. Geographic information systems were used to establish a correlation between environmental features and cardiovascular risk parameters. A total number of 832 (age range 18-74 years) individuals were selected for this study. Body mass index was significantly and positively related to the distance to parks (ρ = 0.079, p < 0.05), but negatively related to the distance to markets (ρ = -0.125, p < 0.05). In addition, waist circumference was similar and positively related to distance to parks (ρ = 0.097, p < 0.05) and negatively related to distance to markets (ρ = -0.092, p < 0.05). With respect to biochemical parameters, when there was an increase in the distance to markets, high-density lipoprotein cholesterol increased and glycemia decreased. The results of this study suggest the importance of the role of environmental factors such as parks and markets in the development of cardiovascular risk.