EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Estimating danofloxacin withdrawal time in broiler chickens based on physiologically based pharmacokinetics modeling.
PMID 25236844
Abstract
In this study, a physiologically based pharmacokinetics (PBPK) model was firstly developed for danofloxacin in healthy broiler chickens after a single oral administration at 5xa0mg/kg bw. Then, the model extrapolation from healthy chickens to those infected with Pasteurella multocidaones was performed. The healthy model was validated through a comparison of predicted and previously published concentrations, which indicated that the healthy PBPK model had good predictive ability in plasma, lung, muscle, liver, and kidney, especially at the later sampling time points. Multiple dosing of administration was incorporated into the healthy and infected models. In addition, a Monte Carlo simulation (MCS) included 1000 iterations was further incorporated into both models to predict the withdrawal times of danofloxacin in healthy and infected chickens, which were estimated to be 3 and 2xa0days, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33700
Danofloxacin, VETRANAL, analytical standard
C19H20FN3O3