EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Streptomyces burgazadensis sp. nov., isolated from soil.
PMID 25237147
Abstract
A novel actinobacterial strain, designated Z1R7(T), was isolated from a soil sample collected from Burgazada, in the Marmara Sea (Turkey), and the strain identity was determined using a polyphasic taxonomic approach. The organism had chemotaxonomic and morphological properties consistent with its classification in the genus Streptomyces and it formed a distinct phyletic line in the 16S rRNA gene tree, together with the type strains Streptomyces specialis GW41-1564(T) (95.76 %), Streptomyces mayteni YIM 60475(T) (95.64 %), Streptomyces hainanensis YIM 47672(T) (95.53 %), Streptomyces hoynatensis S1412(T) (95.29 %), Streptomyces avicenniae MCCC 1A01535(T) (94.74 %), Streptomyces sedi YIM 65188(T) (94.59 %) and Streptomyces zhaozhouensis NEAU-LZS-5(T) (94.68 %). Chomotaxonomic data revealed that strain Z1R7(T) possesed MK-9 (H8) as the predominant menaquinone, ll-diaminopimelic acid as the diagnostic diamino acid, and galactose, glucose and ribose as whole cell sugars. Diphosphatidylglycerol, phoshphatidylethanolamine and phosphatidylinositol were the predominant polar lipids; iso-C16 : 0, anteiso-C17 : 0 and anteiso-C15 : 0 were the major fatty acids, and the genomic DNA G+C content was 69.4 mol%. On the basis of these genotypic and phenotypic data, it is proposed that isolate Z1R7(T) ( = KCTC 29434(T) = DSM 42126(T)) should be classified in the genus Streptomyces as Streptomyces burgazadensis sp. nov.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33240
2,6-Diaminopimelic acid, ≥97.0% (NT)
C7H14N2O4
D1377
2,6-Diaminopimelic acid, ≥98% (TLC)
C7H14N2O4