EMAIL THIS PAGE TO A FRIEND
European journal of oral implantology
Immediate loading of implants placed in patients with untreated periodontal disease: a 5-year prospective cohort study.
PMID 25237673
Abstract
To report the 5-year outcome of immediately loaded dental implants in patients with untreated periodontal disease. This prospective cohort study included 103 consecutive patients (51 females and 52 males) with an average age of 52 years (range: 22 to 80 years) who were rehabilitated with 380 implants supporting 145 prostheses in both jaws (40 single; 33 partial and 72 complete rehabilitations). The implants were inserted in patients with active and untreated periodontitis. The patients did not receive any previous periodontal treatment before implant surgery, except for an oral hygiene session immediately before the implant surgery. In maintenance (every 6 months) patients received periodontal treatment as needed. Outcome measures were: prostheses failures; implant failures; complications; and marginal bone level changes. Fifteen patients dropped out of the study (14.6%). Two patients lost two implants (FDI positions nos.12 and 42), rendering a cumulative survival rate of 97.9% and 99.4% at 5 years of followup using the patient and implant as unit of analysis, respectively. The average (standard deviation) marginal bone resorption was 0.71 mm (0.42 mm) at 5 years. Mechanical complications occurred in 14 patients, consisting of prostheses fractures (10 provisional prostheses and 4 definitive prostheses). Thirteen implants (3.9%) in 13 patients (14.8%) presented peri-implant pathology. Within the limitations of this study, it is possible to conclude that the rehabilitation of patients with untreated periodontitis using immediately loaded dental implants is feasible in the medium-term, when periodontal therapy is provided after rehabilitation and the patients are regularly maintained.