PloS one
Curcumin suppresses crosstalk between colon cancer stem cells and stromal fibroblasts in the tumor microenvironment: potential role of EMT.
PMID 25238234
Abstract
Interaction of stromal and tumor cells plays a dynamic role in initiating and enhancing carcinogenesis. In this study, we investigated the crosstalk between colorectal cancer (CRC) cells with stromal fibroblasts and the anti-cancer effects of curcumin and 5-Fluorouracil (5-FU), especially on cancer stem cell (CSC) survival in a 3D-co-culture model that mimics in vivo tumor microenvironment. Colon carcinoma cells HCT116 and MRC-5 fibroblasts were co-cultured in a monolayer or high density tumor microenvironment model in vitro with/without curcumin and/or 5-FU. Monolayer tumor microenvironment co-cultures supported intensive crosstalk between cancer cells and fibroblasts and enhanced up-regulation of metastatic active adhesion molecules (β1-integrin, ICAM-1), transforming growth factor-β signaling molecules (TGF-β3, p-Smad2), proliferation associated proteins (cyclin D1, Ki-67) and epithelial-to-mesenchymal transition (EMT) factor (vimentin) in HCT116 compared with tumor mono-cultures. High density tumor microenvironment co-cultures synergistically increased tumor-promoting factors (NF-κB, MMP-13), TGF-β3, favored CSC survival (characterized by up-regulation of CD133, CD44, ALDH1) and EMT-factors (increased vimentin and Slug, decreased E-cadherin) in HCT116 compared with high density HCT116 mono-cultures. Interestingly, this synergistic crosstalk was even more pronounced in the presence of 5-FU, but dramatically decreased in the presence of curcumin, inducing biochemical changes to mesenchymal-epithelial transition (MET), thereby sensitizing CSCs to 5-FU treatment. Enrichment of CSCs, remarkable activation of tumor-promoting factors and EMT in high density co-culture highlights that the crosstalk in the tumor microenvironment plays an essential role in tumor development and progression, and this interaction appears to be mediated at least in part by TGF-β and EMT. Modulation of this synergistic crosstalk by curcumin might be a potential therapy for CRC and suppress metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405445
Anti-ALDH1A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002123
Anti-ALDH1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106201
Anti-ALDH1A1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2701626
Anti-aldh1a2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501484
Anti-ALDH2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA051065
Anti-ALDH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410363
Anti-ALDH2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21124 Anti-MMP13 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406136
Anti-MMP13 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2104396
Anti-MMP13 antibody produced in rabbit, affinity isolated antibody
SAB4504207
Anti-phospho-Smad2 (pSer250) antibody produced in rabbit, affinity isolated antibody
SAB4300251
Anti-phospho-SMAD2 (pSer467) antibody produced in rabbit, affinity isolated antibody
SAB4300252
Anti-phospho-SMAD2 (pThr220) antibody produced in rabbit, affinity isolated antibody
WH0000216M1 Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 1A2, purified immunoglobulin, buffered aqueous solution
SAB1403542
Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 1G6, purified immunoglobulin, buffered aqueous solution
SAB5300519 Monoclonal Anti-ALDH1A1 antibody produced in mouse, clone 5A11, ascites fluid
WH0000217M1
Monoclonal Anti-ALDH2 antibody produced in mouse, clone 1E5, purified immunoglobulin, buffered aqueous solution
SAB1408989
Monoclonal Anti-ALDH2 antibody produced in mouse, clone 4F10, purified immunoglobulin, buffered aqueous solution
SAB1408990
Monoclonal Anti-ALDH2 antibody produced in mouse, clone 4C11, purified immunoglobulin, buffered aqueous solution
WH0004322M7
Monoclonal Anti-MMP13 antibody produced in mouse, clone 3B11, purified immunoglobulin, buffered aqueous solution