EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
pH-sensitive chitosan/alginate core-shell nanoparticles for efficient and safe oral insulin delivery.
PMID 25239194
Abstract
Chitosan-alginate (CS/ALG) nanoparticles were prepared by formation of an ionotropic pre-gelation of an alginate (ALG) core entrapping insulin, followed by chitosan (CS) polyelectrolyte complexation, for successful oral insulin administration. Mild preparation process without harsh chemicals is aimed at improving insulin bio-efficiency in in vivo model. The nanoparticles showed an average particle size of 100-200 nm in dynamic light scattering (DLS), with almost spherical or sub-spherical shape and ∼ 85% of insulin encapsulation. Again, retention of almost entire amount of encapsulated insulin in simulated gastric buffer followed by its sustained release in simulated intestinal condition proved its pH sensitivity in in vitro release studies. Significant hypoglycemic effects with improved insulin-relative bioavailability (∼ 8.11%) in in vivo model revealed the efficacy of these core-shell nanoparticles of CS/ALG as an oral insulin carrier. No systemic toxicity was found after its peroral treatment, suggesting these core-shell nanoparticles as a promising device for potential oral insulin delivery.