EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Sildenafil alters retinal function in mouse carriers of retinitis pigmentosa.
PMID 25239397
Abstract
Sildenafil, the active ingredient in Viagra, has been reported to cause transient visual disturbance from inhibition of phosphodiesterase 6 (PDE6), a key enzyme in the visual phototransduction pathway. This study investigated the effects of sildenafil on the rd1(+/-) mouse, a model for carriers of Retinitis Pigmentosa which exhibit normal vision but may have a lower threshold for cellular stress caused by sildenafil due to a heterozygous mutation in PDE6. Sildenafil caused a dose-dependent decrease in electroretinogram (ERG) responses of normal mice which mostly recovered two days post administration. In contrast, rd1(+/-) mice exhibited a significantly reduced photoreceptor and a supernormal bipolar cell response to sildenafil within 1 h of treatment. Carrier mice retinae took two weeks to return to baseline levels suggesting sildenafil has direct effects on both the inner and outer retina and these effects differ significantly between normal and carrier mice. Anatomically, an increase in expression of the early apoptotic marker, cytochrome C in rd1(+/-) mice indicated that the effects of sildenafil on visual function may lead to degeneration. The results of this study are significant considering approximately 1 in 50 people are likely to be carriers of recessive traits leading to retinal degeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001578
Sildenafil citrate, European Pharmacopoeia (EP) Reference Standard
C22H30N6O4S ·C6H8O7
PZ0003
Sildenafil citrate salt, ≥98% (HPLC)
C22H30N6O4S ·C6H8O7