EMAIL THIS PAGE TO A FRIEND
FEBS letters
Saturated fatty acid-induced miR-195 impairs insulin signaling and glycogen metabolism in HepG2 cells.
PMID 25240198
Abstract
MicroRNAs (miRNAs) play an important role in insulin signaling and insulin secretion, but the role of miRNAs in the association between obesity and hepatic insulin resistance is largely unknown. This study reports that saturated fatty acid (SFA) and high fat diet (HFD) significantly induce miR-195 expression in hepatocytes, and that the insulin receptor (INSR), not insulin receptor substrate-1 (IRS-1), is a direct target of miR-195. Furthermore, the ectopic expression of miR-195 suppresses the expression of INSR, thereby impairing the insulin signaling cascade and glycogen synthesis in HepG2 cells. These findings suggest that the dysregulation of miR-195 by SFA is a detrimental factor for hepatic insulin sensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7153 Anti-Insulin Receptor Substrate 1 (IRS-1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500281
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501568
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501569
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501570
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501571
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501572
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501573
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4300482
Anti-IRS1 (Ab-636) antibody produced in rabbit, affinity isolated antibody
SAB4300483
Anti-IRS1 (Ab-639) antibody produced in rabbit, affinity isolated antibody
SAB1406045
Anti-IRS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4503887
Anti-phospho-IRS-1 (pSer1101) antibody produced in rabbit, affinity isolated antibody
SAB4504442
Anti-phospho-IRS-1 (pSer307) antibody produced in rabbit, affinity isolated antibody
SAB4300653
Anti-phospho-IRS-1 (pSer312) antibody produced in rabbit, affinity isolated antibody
SAB4504443
Anti-phospho-IRS-1 (pSer312) antibody produced in rabbit, affinity isolated antibody
SAB4503927
Anti-phospho-IRS-1 (pSer323) antibody produced in rabbit, affinity isolated antibody
SAB4503888
Anti-phospho-IRS-1 (pSer612) antibody produced in rabbit, affinity isolated antibody
SAB4504444
Anti-phospho-IRS-1 (pSer636) antibody produced in rabbit, affinity isolated antibody
SAB4504032
Anti-phospho-IRS-1 (pSer794) antibody produced in rabbit, affinity isolated antibody
SAB4504661
Anti-phospho-IRS-1 (pTyr896) antibody produced in rabbit, affinity isolated antibody
SAB4300185
Anti-phospho-IRS1 (pSer636) antibody produced in rabbit, affinity isolated antibody
SAB4300186
Anti-phospho-IRS1 (pSer639) antibody produced in rabbit, affinity isolated antibody