EMAIL THIS PAGE TO A FRIEND
Pharmacology, biochemistry, and behavior
The role of alpha-2 adrenoceptors in the anticonvulsant effects of adenosine on pentylenetetrazole-induced seizure threshold in mice.
PMID 25242809
Abstract
Adenosine has anticonvulsant effects in various models of seizures. Alpha-2 adrenoceptors have also demonstrated different effects in different models of epilepsy. In this study, the role of alpha-2 adrenoceptors in the anticonvulsant effects of adenosine in mice was determined according to the method of intravenous pentylenetetrazole-induced seizure. In this study, N(6)-cyclohexyladenosine (CHA) (a selective A1 receptor agonist), clonidine (an alpha-2 adrenoceptors agonist), yohimbine (an alpha-2 adrenoceptors antagonist) and 8-cyclopentyl-1,3-dimethylxanthine (8-CPT) (a selective A1 receptor antagonist) were used. CHA at doses of 0.5, 1 and 2mg/kg significantly increased seizure threshold with the maximum anticonvulsant effect at 2mg/kg. Yohimbine (0.1, 1 and 10mg/kg), clonidine (0.1, 0.5, 1 and 2mg/kg) and 8-CPT (0.5, 1, 2 and 4mg/kg) had no effect on seizure by itself. Combination of yohimbine (10mg/kg) and CHA (0.25mg/kg) increased clonic seizure latency showing that yohimbine and CHA have an additive effect. Increasing the seizure threshold created by combining ineffective doses of yohimbine (10mg/kg) and CHA (0.25mg/kg) was completely inhibited by 8-CPT (4mg/kg) or clonidine (1 and 2mg/kg). Clonidine (0.5, 1 and 2mg/kg) inhibited the anticonvulsant effects of CHA (2mg/kg). Combination of 8-CPT (1mg/kg) and clonidine (0.5mg/kg) which completely inhibited the anticonvulsant effect of CHA (2mg/kg) indicates that 8-CPT and clonidine have an additive effect. In conclusion, adenosine and yohimbine exhibit an additive effect on the enhancement of the pentylenetetrazole-induced seizure threshold in mice, indicating the interaction of alpha-2 adrenoceptors and A1 adenosine receptors.
Related Materials