EMAIL THIS PAGE TO A FRIEND
BioMed research international
Epinephrine enhances the response of macrophages under LPS stimulation.
PMID 25243125
Abstract
Trauma associated with infection may directly trigger a neuroendocrine reaction in vivo while the hormone epinephrine is known to mediate immune responses to inflammation after injury. However, the role of epinephrine during the earliest stage of trauma still remains unclear. We therefore explored the role of epinephrine on activated macrophages under LPS stimulation in vitro as well as the mechanisms underlying its effect. Dose- and time-dependent effects of epinephrine on macrophage immune function were assessed after LPS activation. We also employed CD14 siRNA interference to investigate whether CD14 played a role in the mechanism underlying the effect of epinephrine on LPS-induced macrophage responses. Our results showed that epinephrine pretreatment (10 ng/mL) significantly promoted immune responses from LPS stimulated macrophages, including phagocytic rate, phagocytic index, TNFα/IL-1β/IL-10 secretion, and CD14 expression (P < 0.05). Moreover, TNFα/IL-1β/IL-10 levels attained their peak value 1 hour after incubation with 10 ng/mL epinephrine (P < 0.05), and CD14 siRNA transfection dramatically decreased phagocytosis and cytokine secretion by LPS-activated macrophages (P < 0.05). We therefore conclude that 10 ng/mL epinephrine enhances immune responses from macrophages under LPS stimulation and that the underlying mechanism may relate to CD14 upregulation on the surface of macrophages.