EMAIL THIS PAGE TO A FRIEND
Progress in neuro-psychopharmacology & biological psychiatry
Clozapine induces chloride channel-4 expression through PKA activation and modulates CDK5 expression in SH-SY5Y and U87 cells.
PMID 25246152
Abstract
Second-generation antipsychotic drugs, such as clozapine, were reported to enhance neurite outgrowth by nerve growth factor in PC12 cells. The authors previously showed that chloride channel 4 (CLC-4) is responsible for nerve growth factor-induced neurite outgrowth in neuronal cells. In this study, we examined whether clozapine induces CLC-4 in neuroblastoma and glioma cells. The effect of clozapine on CLC-4 expression was examined in neuroblastoma (SH-SY5Y) and glioma (U87) cells. To investigate the signaling pathway responsible for clozapine-induced CLC-4 expression, the phosphorylation of cAMP response element-binding protein (CREB), which binds CRE in the promoter of the human CLC-4 gene, was examined. To identify the target of clozapine induced CLC-4, CLC-4 siRNA was introduced to neuroblastoma and glioma cells for functional knockdown. We observed that clozapine increased CLC-4 expression in both SH-SY5Y and U87 cells. Clozapine induced CREB phosphorylation, but in the presence of inhibitor of protein kinase A (an upstream kinase of CREB) clozapine-induced CLC-4 expression was suppressed. Finally, we found that CLC-4 knockdown suppressed clozapine-induced cyclin-dependent kinase 5 (CDK5) expression in SH-SY5Y and U-87 cells suggesting CDK5 as potential molecular target of clozapine induced CLC-4 expression. The results of the present study suggest that clozapine's therapeutic effect may include the induction of CLC-4 which is dependent on CREB activation via PKA. We also found that functional knockdown of CLC-4 resulted in reduction of CDK5 expression, which may also be implicated in clozapine's therapeutic effect.