EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Functional avidity-driven activation-induced cell death shapes CTL immunodominance.
PMID 25246498
Abstract
Immunodominance is a complex phenomenon that relies on a mere numerical concept, while being potentially influenced at every step of the immune response. We investigated the mechanisms leading to the establishment of CTL immunodominance in a retroviral model and found that the previously defined subdominant Env-specific CD8(+) T cells are endowed with an unexpectedly higher functional avidity than is the immunodominant Gag-recognizing counterpart. This high avidity, along with the Env Ag overload, results in a supraoptimal TCR engagement. The overstimulation makes Env-specific T lymphocytes more susceptible to apoptosis, thus hampering their expansion and leading to an unintentional "immune kamikazing." Therefore, Ag-dependent, hyperactivation-induced cell death can be regarded as a novel mechanism in the establishment of the immunodominance that restrains and opposes the expansion of high-avidity T cells in favor of lower-affinity populations.