EMAIL THIS PAGE TO A FRIEND
PLoS biology
Haploinsufficiency of Dmxl2, encoding a synaptic protein, causes infertility associated with a loss of GnRH neurons in mouse.
PMID 25248098
Abstract
Characterization of the genetic defects causing gonadotropic deficiency has made a major contribution to elucidation of the fundamental role of Kisspeptins and Neurokinin B in puberty onset and reproduction. The absence of puberty may also reveal neurodevelopmental disorders caused by molecular defects in various cellular pathways. Investigations of these neurodevelopmental disorders may provide information about the neuronal processes controlling puberty onset and reproductive capacity. We describe here a new syndrome observed in three brothers, which involves gonadotropic axis deficiency, central hypothyroidism, peripheral demyelinating sensorimotor polyneuropathy, mental retardation, and profound hypoglycemia, progressing to nonautoimmune insulin-dependent diabetes mellitus. High-throughput sequencing revealed a homozygous in-frame deletion of 15 nucleotides in DMXL2 in all three affected patients. This homozygous deletion was associated with lower DMXL2 mRNA levels in the blood lymphocytes of the patients. DMXL2 encodes the synaptic protein rabconnectin-3α, which has been identified as a putative scaffold protein for Rab3-GAP and Rab3-GEP, two regulators of the GTPase Rab3a. We found that rabconnectin-3α was expressed in exocytosis vesicles in gonadotropin-releasing hormone (GnRH) axonal extremities in the median eminence of the hypothalamus. It was also specifically expressed in cells expressing luteinizing hormone (LH) and follicle-stimulating hormone (FSH) within the pituitary. The conditional heterozygous deletion of Dmxl2 from mouse neurons delayed puberty and resulted in very low fertility. This reproductive phenotype was associated with a lower number of GnRH neurons in the hypothalamus of adult mice. Finally, Dmxl2 knockdown in an insulin-secreting cell line showed that rabconnectin-3α controlled the constitutive and glucose-induced secretion of insulin. In conclusion, this study shows that low levels of DMXL2 expression cause a complex neurological phenotype, with abnormal glucose metabolism and gonadotropic axis deficiency due to a loss of GnRH neurons. Our findings identify rabconectin-3α as a key controller of neuronal and endocrine homeostatic processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2102000
Anti-RHCE antibody produced in rabbit, affinity isolated antibody
SAB1302678
ANTI-RHD (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1412742
Anti-RHD antibody produced in mouse, purified immunoglobulin, 1 mg/mL
SAB2107753
Anti-RHD antibody produced in rabbit, affinity isolated antibody
SAB2107754
Anti-RHD antibody produced in rabbit, affinity isolated antibody
L1898
[D-Ala6]-LH-RH acetate salt hydrate, ≥97% (HPLC)
C56H77N17O13 · xC2H4O2 · yH2O
L4663 [D-Ser4]-LH-RH, >97% (HPLC)
C55H75N17O13
L4788
[D-Trp6]-LH-RH-Leu-Arg-Pro-Gly-NH2, ≥96% (HPLC)
C83H115N25O17
49159
D-(+)-Glucose monohydrate, for microbiology, ≥99.0%
C6H12O6 · H2O
16301
D-(+)-Glucose monohydrate, meets analytical specification of Ph. Eur., BP, Ph Franç., 7.0-9.5% water(Karl Fischer)
C6H12O6 · H2O
49161
D-(+)-Glucose monohydrate, tested according to Ph.Eur.
C6H12O6 · H2O
L3538 [D-His(benzyl)6]-LH-RH Fragment 3-9 ethylamide trifluoroacetate salt, >99% (HPLC)
C55H74N14O9
L4538 [D-His(Bzl)6]-LH-RH Fragment 1-7, >96% (HPLC)
C53H62N12O11
L3413 [D-His(Bzl)6]-LH-RH Fragment 4-9 ethylamide trifluoroacetate salt, >95% (HPLC)
C44H64N12O8
L3288 [D-His(Bzl)6]-LH-RH Fragment 5-9 Ethylamide trifluoroacetate salt, >98% (HPLC)
C41H59N11O6
L4513
[des-Gly10, D-Ala6]-LH-RH ethylamide acetate salt hydrate, ≥97% (HPLC), powder
C56H78N16O12 · xC2H4O2 · yH2O
L2761
[des-Gly10, D-His(Bzl)6]-LH-RH ethylamide, ≥97% (HPLC), powder
C66H86N18O12
L4163 [des-Gly10, D-His2, D-Trp6, Pro9]-LH-RH ethylamide trifluoroacetate salt, >98% (HPLC)
C64H83N17O12
L4038 [des-Gly10, D-Ser4, D-Trp6, Pro9]-LH-RH ethylamide trifluoroacetate salt, >98% (HPLC)
C64H83N17O12
L3788 [des-Gly10, D-Tyr5, D-Trp6, Pro9]-LH-RH ethylamide trifluoroacetate salt, >99% (HPLC)
C64H83N17O12
D9559
Dextrose monohydrate, meets USP testing specifications
C6H12O6 · H2O
L5413
Formyl-[D-Trp6]-LH-RH Fragment 2-10, >97% (HPLC)
C60H77N17O12
L4913 [His(3-Methyl)2]-LH-RH, >96% (HPLC)
C56H77N17O13
L0399 Leuprolide acetate salt, ≥98% (HPLC)
C59H84N16O12 · xC2H4O2
L8008
Luteinizing hormone releasing hormone, ≥97% (HPLC)
C55H74N16O14
L4897
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
C60H73N15O13
EHU036851 MISSION® esiRNA, esiRNA human NTS (esiRNA1)
EHU023471 MISSION® esiRNA, esiRNA human NT5E (esiRNA1)
SIHP0370 MISSION® Human Phosphatase, NT5E siRNA1, Nano Scale 250 pmol
SIHP0371 MISSION® Human Phosphatase, NT5E siRNA2, Nano Scale 250 pmol