EMAIL THIS PAGE TO A FRIEND
Updates in surgery
Watch and wait plus intra-operative lymphatic mapping as a novel approach for the surgical removal of a persistent groin lymphocele.
PMID 25248314
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
198218
Patent Blue VF, Dye content 50 %
C27H31N2NaO6S2
76357
Patent blue VF sodium salt, for microscopy
C27H31N2NaO6S2