EMAIL THIS PAGE TO A FRIEND
Mucosal immunology
Reciprocal regulation of lymphoid tissue development in the large intestine by IL-25 and IL-23.
PMID 25249168
Abstract
Isolated lymphoid follicles (ILFs) develop after birth in the small and large intestines (SI and LI) and represent a dynamic response of the gut immune system to the microbiota. Despite their similarities, ILF development in the SI and LI differs on a number of levels. We show that unlike ILF in the SI, the microbiota inhibits ILF development in the colon as conventionalization of germ-free mice reduced colonic ILFs. From this, we identified a novel mechanism regulating colonic ILF development through the action of interleukin (IL)-25 on IL-23 and its ability to modulate T regulatory cell (Treg) differentiation. Colonic ILF develop in the absence of a number of factors required for the development of their SI counterparts and can be specifically suppressed by factors other than IL-25. However, IL-23 is the only factor identified that specifically promotes colonic ILFs without affecting SI-ILF development. Both IL-23 and ILFs are associated with inflammatory bowel disease, suggesting that disruption to this pathway may have an important role in the breakdown of microbiota-immune homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA042455 Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA043579
Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA049348
Anti-CR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501275
Anti-THY1 antibody produced in goat, affinity isolated antibody
HPA003733
Anti-THY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700245
Monoclonal Anti-CD21 antibody produced in mouse, clone LT21, purified immunoglobulin, buffered aqueous solution
WH0007070M1
Monoclonal Anti-THY1 antibody produced in mouse, clone 3F9, purified immunoglobulin, buffered aqueous solution
SAB4200497
Monoclonal Anti-THY1 antibody produced in mouse, ~1.0 mg/mL, purified immunoglobulin
AMAB90846
Monoclonal Anti-THY1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1040, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90844
Monoclonal Anti-THY1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1028, purified immunoglobulin, buffered aqueous glycerol solution