EMAIL THIS PAGE TO A FRIEND
Biology of reproduction
Transcriptome profiling of granulosa and theca cells during dominant follicle development in the horse.
PMID 25253738
Abstract
Several aspects of equine ovarian physiology are unique among domestic species. Moreover, follicular growth patterns are very similar between horses and humans. This study aimed to characterize, for the first time, global gene expression profiles associated with growth and preovulatory (PO) maturation of equine dominant follicles. Granulosa cells (GCs) and theca interna cells (TCs) were harvested from follicles (n = 5) at different stages of an ovulatory wave in mares corresponding to early dominance (ED; diameter ≥22 mm), late dominance (LD; ≥33 mm) and PO stage (34 h after administration of crude equine gonadotropins at LD stage), and separately analyzed on a horse gene expression microarray, followed by validation using quantitative PCR and immunoblotting/immunohistochemistry. Numbers of differentially expressed transcripts (DETs; ≥2-fold; P < 0.05) during the ED-LD and LD-PO transitions were 546 and 2419 in GCs and 5 and 582 in TCs. The most prominent change in GCs was the down-regulation of transcripts associated with cell division during both ED-LD and LD-PO. In addition, DET sets during LD-PO in GCs were enriched for genes involved in cell communication/adhesion, antioxidation/detoxification, immunity/inflammation, and cholesterol biosynthesis. In contrast, the largest change in TCs during the LD-PO transition was an up-regulation of genes involved in immune activation, with other DET sets mapping to GPCR/cAMP signaling, lipid/amino acid metabolism, and cell proliferation/survival and differentiation. In conclusion, distinct expression profiles were identified between growing and PO follicles and, particularly, between GCs and TCs within each stage. Several DETs were identified that have not been associated with follicle development in other species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2229 Anti-Apolipoprotein J antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502026
Anti-Clu antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502095
Anti-CLU antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA000572
Anti-CLU antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500199
Anti-Clusterin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501425
Anti-DAG1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2106735
Anti-DAG1 antibody produced in rabbit, affinity isolated antibody
AV34376
Anti-ENO1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
GW22683 Anti-ENO1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV34377
Anti-ENO1 antibody produced in rabbit, affinity isolated antibody
SAB2106422
Anti-ENO1 antibody produced in rabbit, affinity isolated antibody
SAB1401216
Anti-INHBB antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA035386
Anti-INHBB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2900220
Anti-ITGB1 (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900236
Anti-ITGB1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107342
Anti-ITGB1 antibody produced in rabbit, affinity isolated antibody
AV100955
Anti-MBP-1 antibody produced in rabbit, affinity isolated antibody
SAB4700394 Monoclonal Anti-CD29 antibody produced in mouse, clone MEM-101A, purified immunoglobulin, buffered aqueous solution
WH0001191M1
Monoclonal Anti-CLU antibody produced in mouse, clone 1A11, purified immunoglobulin, buffered aqueous solution
WH0001605M1
Monoclonal Anti-DAG1 antibody produced in mouse, clone 2A3, purified immunoglobulin, buffered aqueous solution
WH0002023M1
Monoclonal Anti-ENO1 antibody produced in mouse, clone 8G8, purified immunoglobulin, buffered aqueous solution
SAB1403772
Monoclonal Anti-ENO1 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
P8891
Propantheline bromide, ≥97% (TLC), powder
C23H30BrNO3
1570508
Propantheline bromide, United States Pharmacopeia (USP) Reference Standard
C23H30BrNO3