EMAIL THIS PAGE TO A FRIEND
European journal of cancer prevention : the official journal of the European Cancer Prevention Organisation (ECP)
Resveratrol enhances the chemopreventive effect of celecoxib in chemically induced breast cancer in rats.
PMID 25254309
Abstract
Resveratrol and celecoxib were used as chemopreventive agents in animal models of carcinogenesis, and exert antiproliferative and proapoptotic effects on cancer cells. Therefore, the aim of this study was to evaluate whether combining resveratrol with celecoxib may exert more potent anticarcinogenic effects than the single agents. Mammary carcinogenesis was initiated in 70 female Sprague-Dawley rats with N-methyl-N-nitrosourea (NMU). The chemoprevention with resveratrol, celecoxib, and their combination started 2 weeks before the first carcinogen dose and lasted until the end of the experiment. Tumor incidence and frequency, latency period, tumor volume, the expression of cyclooxygenase 2 (COX2) and growth differentiation factor 15 (GDF15), and also the formation of reactive oxygen species were analyzed using different methods. In addition, the levels of resveratrol and its metabolites in blood and selected tumor tissues were determined by high-performance liquid chromatography. Finally, the anticancer effects of the reagents were studied in the human breast cancer cell line MCF-7. Celecoxib as a single agent significantly decreased tumor frequency, prolonged tumor latency, and decreased the total number of malignant tumors compared with the NMU conditions. Tumor volume was nonsignificantly reduced (0.68±0.25 vs. 0.93±0.28 cm3). Importantly, the addition of resveratrol to celecoxib reduced tumor volume by 60% compared with celecoxib alone (from 0.68±0.25 to 0.27±0.07 cm3, P<0.05). Furthermore, the combination of resveratrol and celecoxib reduced tumor frequency by 29% compared with celecoxib alone (P=0.53). Tumor latency was not influenced by this combination compared with celecoxib alone (126.56±3.45 vs. 120.71±4.08 days). In addition, COX2 mRNA and immunoreactive protein stained on tumor sections were reduced and GDF15 protein increased significantly by the combination studied compared with the NMU conditions. In agreement with these data, a significant reduction in reactive oxygen species in blood lymphocytes of the combination was detected, which may have contributed toward the cancer-preventive effects of this application. This study showed that in NMU-induced mammary cancer in rats, the combination of resveratrol and celecoxib led to a significant reduction in all tumor parameters. In addition, in terms of tumor volume, the combination was more efficient than celecoxib as a single agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1517
N-Nitroso-N-methylurea, ISOPAC®
C2H5N3O2
N4766
N-Nitroso-N-methylurea, Bulk package
C2H5N3O2