EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Beyond C, H, O, and N! Analysis of the elemental composition of U.S. FDA approved drug architectures.
PMID 25255063
Abstract
The diversity of elements among U.S. Food and Drug Administration (FDA) approved pharmaceuticals is analyzed and reported, with a focus on atoms other than carbon, hydrogen, oxygen, and nitrogen. Our analysis reveals that sulfur, chlorine, fluorine, and phosphorous represent about 90% of elemental substitutions, with sulfur being the fifth most used element followed closely by chlorine, then fluorine and finally phosphorous in the eighth place. The remaining 10% of substitutions are represented by 16 other elements of which bromine, iodine, and iron occur most frequently. The most detailed parts of our analysis are focused on chlorinated drugs as a function of approval date, disease condition, chlorine attachment, and structure. To better aid our chlorine drug analyses, a new poster showcasing the structures of chlorinated pharmaceuticals was created specifically for this study. Phosphorus, bromine, and iodine containing drugs are analyzed closely as well, followed by a discussion about other elements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
22754
Chlorine, puriss., ≥99.8%
Cl2