EMAIL THIS PAGE TO A FRIEND
eLife
The neural crest is a source of mesenchymal stem cells with specialized hematopoietic stem cell niche function.
PMID 25255216
Abstract
Mesenchymal stem cells (MSCs) and osteolineage cells contribute to the hematopoietic stem cell (HSC) niche in the bone marrow of long bones. However, their developmental relationships remain unclear. In this study, we demonstrate that different MSC populations in the developing marrow of long bones have distinct functions. Proliferative mesoderm-derived nestin(-) MSCs participate in fetal skeletogenesis and lose MSC activity soon after birth. In contrast, quiescent neural crest-derived nestin(+) cells preserve MSC activity, but do not generate fetal chondrocytes. Instead, they differentiate into HSC niche-forming MSCs, helping to establish the HSC niche by secreting Cxcl12. Perineural migration of these cells to the bone marrow requires the ErbB3 receptor. The neonatal Nestin-GFP(+) Pdgfrα(-) cell population also contains Schwann cell precursors, but does not comprise mature Schwann cells. Thus, in the developing bone marrow HSC niche-forming MSCs share a common origin with sympathetic peripheral neurons and glial cells, and ontogenically distinct MSCs have non-overlapping functions in endochondrogenesis and HSC niche formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA011753
Anti-CAMK4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA017206
Anti-CAMK4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37025
Anti-CAMK4 antibody produced in rabbit, IgG fraction of antiserum
SAB4503233
Anti-CAMK4 antibody produced in rabbit, affinity isolated antibody
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV49121
Anti-ST6GALNAC4 antibody produced in rabbit, affinity isolated antibody
SAB2701445
Anti-ST6GALNAC4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103892
Anti-TH, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2103911
Anti-TH, (N-terminal) antibody produced in rabbit, affinity isolated antibody
T8700 Anti-Tyrosine Hydroxylase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots
WH0000059M2
Monoclonal Anti-ACTA2 antibody produced in mouse, clone 4A8-2H3, purified immunoglobulin, buffered aqueous solution
SAB1403519
Monoclonal Anti-ACTA2 antibody produced in mouse, clone 1D11-2B9, purified immunoglobulin, buffered aqueous solution