EMAIL THIS PAGE TO A FRIEND
Experimental dermatology
Egr-1 is a key regulator of IL-17A-induced psoriasin upregulation in psoriasis.
PMID 25256120
Abstract
The early growth response (Egr)-1 is a transcriptional factor which plays an important role in the regulation of cell growth, differentiation, cell survival and immune responses. Emerging evidences including our data demonstrate that the Egr-1 expression is up-regulated in the psoriatic skin lesions. The purpose of this study was to investigate the significance and regulatory mechanism of Egr-1 in the pathogenesis of psoriasis. Through microarray analysis, we found out that psoriasin (S100A7) expression was increased in the Egr-1 overexpressed cells. Our results showed that IL-17A increased Egr-1 expression in the skin of psoriatic patients and cultured human keratinocytes. We then investigated activation of mitogen-activated protein kinase as an upstream signal regulator of Egr-1 expression. IL-17A-induced Egr-1 expression was suppressed by ERK inhibitor. In addition, IL-17A induced psoriasin expression in cultured keratinocytes and the skin of IL-17A intradermally injected mouse. IL-17A-mediated psoriasin upregulation was reduced after treatment of small interfering RNAs against Egr-1. Furthermore, the results of chromatin immunoprecipitation assays demonstrated that Egr-1 directly binds the psoriasin promoter. Our findings present a novel signalling mechanism by which IL-17A can induce the Egr-1-dependent psoriasin expression via the ERK pathway in human keratinocytes. This study suggests that Egr-1 may be a novel and important modulator in IL-17A-mediated immune response in psoriasis.