EMAIL THIS PAGE TO A FRIEND
The EMBO journal
SNW1 enables sister chromatid cohesion by mediating the splicing of sororin and APC2 pre-mRNAs.
PMID 25257309
Abstract
Although splicing is essential for the expression of most eukaryotic genes, inactivation of splicing factors causes specific defects in mitosis. The molecular cause of this defect is unknown. Here, we show that the spliceosome subunits SNW1 and PRPF8 are essential for sister chromatid cohesion in human cells. A transcriptome-wide analysis revealed that SNW1 or PRPF8 depletion affects the splicing of specific introns in a subset of pre-mRNAs, including pre-mRNAs encoding the cohesion protein sororin and the APC/C subunit APC2. SNW1 depletion causes cohesion defects predominantly by reducing sororin levels, which causes destabilisation of cohesin on DNA. SNW1 depletion also reduces APC/C activity and contributes to cohesion defects indirectly by delaying mitosis and causing "cohesion fatigue". Simultaneous expression of sororin and APC2 from intron-less cDNAs restores cohesion in SNW1-depleted cells. These results indicate that the spliceosome is required for mitosis because it enables expression of genes essential for cohesion. Our transcriptome-wide identification of retained introns in SNW1- and PRPF8-depleted cells may help to understand the aetiology of diseases associated with splicing defects, such as retinosa pigmentosum and cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA017370
Anti-SNW1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
791733
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3KO2
P1190
Potassium acetate, for molecular biology, ≥99.0%
C2H3KO2
P5708
Potassium acetate, BioXtra, ≥99.0%
C2H3KO2
P1222
Potassium acetate, meets USP testing specifications
C2H3KO2
255785
Potassium acetate, 99.98% trace metals basis, ~100 ppm Rb, <50 ppm Cs
C2H3KO2
236497
Potassium acetate, ACS reagent, ≥99.0%
C2H3KO2
P1147
Potassium acetate, ReagentPlus®, ≥99.0%
C2H3KO2
60035
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
C2H3KO2
32309
Potassium acetate, puriss. p.a., ≥99% (perchloric acid titration)
C2H3KO2
25059
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
C2H3KO2
W292001
Potassium acetate, 98%
C2H3KO2
95843
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
C2H3KO2
ARK2161
Sodium butyrate, PharmaGrade, USP/NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C4H7NaO2
07596
Sodium butyrate, certified reference material, TraceCERT®
C4H7NaO2
B5887
Sodium butyrate, ≥98.5% (GC)
C4H7NaO2
19364
Sodium butyrate, ≥99.0% (GC)
C4H7NaO2
303410
Sodium butyrate, 98%
C4H7NaO2