EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
An analysis of the black crusts from the Seville Cathedral: a challenge to deepen the understanding of the relationships among microstructure, microchemical features and pollution sources.
PMID 25260161
Abstract
The Cathedral of Seville is one of the most important buildings in the whole of southern Spain. It suffers, like most of the historical buildings located in urban environments, from several degradation phenomena related to the high pollution level. Undoubtedly, the formation of black crusts plays a crucial role in the decay of the stone materials belonging to the church. Their formation occurs mainly on carbonate building materials, whose interaction with a sulfur oxide-enriched atmosphere leads to the transformation of calcium carbonate (calcite) into calcium sulfate dihydrate (gypsum) which, together with embedded carbonaceous particles, forms the black crusts on the stone surface. To better understand the composition and the formation dynamics of this degradation product and to identify the pollutant sources and evaluate their impact on the stone material, an analytical study was carried out on the black crust samples collected from different areas of the building. For a complete characterization of the black crusts, several techniques were used, including laser ablation inductively coupled plasma mass spectrometry, Fourier transform infrared spectroscopy, micro infrared spectroscopy, optical and scanning electron microscopy. This battery of tests provided information about the nature and distribution of the mineralogical phases and the elements within the crusts and the crust-substrate interface, contributing to the identification of the major pollution sources responsible for the deterioration of the monument over time. In addition, the results revealed a relation among the height of sampling, the surface exposure and the concentration of heavy metals. Finally, information has been provided about the origin of the concentration gradients of some metals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6763
Calcium carbonate, ReagentPlus®
CCaO3
239216
Calcium carbonate, ACS reagent, ≥99.0%, powder
CCaO3
12010
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
CCaO3
900500
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
CCaO3
238961
Calcium sulfate, with indicator, 4 mesh
CaO4S
456071
Calcium sulfate, with indicator, 6 mesh
CaO4S
238988
Calcium sulfate, with indicator, 8 mesh
CaO4S
238910
Calcium sulfate, without indicator, 4 mesh
CaO4S
238937
Calcium sulfate, without indicator, 8 mesh
CaO4S
255696
Calcium sulfate, ≥99.99% trace metals basis
CaO4S
237132
Calcium sulfate, −325 mesh, 99%
CaO4S
C3771
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
CaO4S · 2H2O
C3210
Calcium sulfate dihydrate, BioXtra, ≥98.0%
CaO4S · 2H2O
12056
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
CaO4S · 2H2O
255548
Calcium sulfate dihydrate, ACS reagent, 98%
CaO4S · 2H2O
12090
Calcium sulfate hemihydrate, purum, ≥97.0%
CaO4S · 0.5H2O
307661
Calcium sulfate hemihydrate, 98%
CaO4S · 0.5H2O
05-1080
Calcium sulfate hemihydrate, CP, calcined
CaO4S · 0.5H2O
238929
Drierite, without indicator, 6 mesh
CaO4S
238945
Drierite, without indicator, 10-20 mesh
CaO4S
737828
Drierite, with indicator, 10-20 mesh
CaO4S