EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Temporal trends in organic carbon content in the main Swiss rivers, 1974-2010.
PMID 25260166
Abstract
Increases in dissolved organic carbon (DOC) concentrations have often been reported in rivers and lakes of the Northern Hemisphere over the last few decades. High-quality organic carbon (OC) concentration data have been used to study the change in DOC and total (TOC) organic carbon concentrations in the main rivers of Switzerland (Rhône, Rhine, Thur and Aar) between 1974 and 2010. These rivers are characterized by high discharge regimes (due to their Alpine origin) and by running in populated areas. Small long term trends (a general statistically significant decrease in TOC and a less clear increase in DOC concentrations), on the order of 1% of mean OC concentration per year, have been observed. An upward trend before 1999 reversed direction to a more marked downward trend from 1999 to 2010. Of the potential causes of OC temporal variation analysed (water temperature, dissolved reactive phosphorus and river discharge), only discharge explains a significant, albeit still small, part of TOC variability (8-31%), while accounting for barely 2.5% of DOC variability. Estimated anthropogenic TOC and DOC loads (treated sewage) to the rivers could account for a maximum of 4-20% of the temporal trends. Such low predictability is a good example of the limitations faced when studying causality and drivers behind small variations in complex systems. River export of OC from Switzerland has decreased significantly over the period. Since about 5.5% of estimated NEP of Switzerland is exported by the rivers, riverine OC fluxes should be taken into account in a detailed carbon budget of the country.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2764
Activated charcoal, untreated, granular, ≤5 mm
C
C2889
Activated charcoal, untreated, granular, 8-20 mesh
C
161551
Activated charcoal, powder, -100 particle size (mesh), decolorizing
C
292591
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
C
242233
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
C
242241
Activated charcoal, DARCO®, 12-20 mesh, granular
C
242268
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
C
242276
Activated charcoal, DARCO®, −100 mesh particle size, powder
C
329428
Activated charcoal, NORIT® ROW 0.8 SUPRA, pellets
C
C3014
Activated charcoal, untreated, granular, 20-60 mesh
C
C3345
Activated charcoal, untreated powder, 100-400 mesh
C
C4386
Activated charcoal, acid-washed with hydrochloric acid
C
C5510
Activated charcoal, acid-washed with phosphoric and sulfuric acids
C
675326
Activated charcoal, DARCO® KB-G
C
675342
Activated charcoal, DARCO® KB-WJ
C
C7606
Activated charcoal, meets USP testing specifications
C
97876
Activated Charcoal Norit®, Norit CA1, from wood, chemically activated, powder
C
37771
Activated Charcoal Norit®, Norit GAC 1240W, from coal, for potable water processing, steam activated, granular
C
22874
Activated Charcoal Norit®, Norit PK 1-3, from peat, steam activated, granular
C
29204
Activated Charcoal Norit®, Norit RB3, for gas purification, steam activated, rod
C
51127
Activated Charcoal Norit®, Norit RB4C, steam activated, rod, for the purification of gas
C
53663
Activated Charcoal Norit®, Norit SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
C
93067
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
C
484164 Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
C
633100 Carbon, nanopowder, <100 nm particle size (TEM)
C
GF29093058
Carbon, chopped fiber, 100g, nominal diameter 0.007mm, fiber length 6mm, grade 34-700
C
GF02887342
Carbon, fabric, 1000x1000mm, weight.muü 375g, thickness 0.4mm, ends x picks/10cm 47x47, 2/2 twill weave
C
GF33494957
Carbon, fabric, 1000x1000mm, weight.muü 92g, thickness 0.15mm, ends x picks/10cm 69x69, plain weave
C
GF07091861
Carbon, fabric, 150x150mm, weight.muü 375g, thickness 0.4mm, ends x picks/10cm 47x47, 2/2 twill weave
C
GF39337724
Carbon, fabric, 250x250mm, weight.muü 92g, thickness 0.15mm, ends x picks/10cm 69x69, plain weave
C