EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Sofosbuvir and ribavirin prevent recurrence of HCV infection after liver transplantation: an open-label study.
PMID 25261839
Abstract
Patients with detectable hepatitis C virus (HCV) RNA at the time of liver transplantation universally experience recurrent HCV infection. Antiviral treatment before transplantation can prevent HCV recurrence, but existing interferon-based regimens are poorly tolerated and are either ineffective or contraindicated in most patients. We performed a trial to determine whether sofosbuvir and ribavirin treatment before liver transplantation could prevent HCV recurrence afterward. In a phase 2, open-label study, 61 patients with HCV of any genotype and cirrhosis (Child-Turcotte-Pugh score, ≤7) who were on waitlists for liver transplantation for hepatocellular carcinoma, received up to 48 weeks of sofosbuvir (400 mg) and ribavirin before liver transplantation. The primary end point was the proportion of patients with HCV-RNA levels less than 25 IU/mL at 12 weeks after transplantation among patients with this HCV-RNA level at their last measurement before transplantation. Sixty-one patients received sofosbuvir and ribavirin, and 46 received transplanted livers. The per-protocol efficacy population consisted of 43 patients who had HCV-RNA level less than 25 IU/mL at the time of transplantation. Of these 43 patients, 30 (70%) had a post-transplantation virologic response at 12 weeks, 10 (23%) had recurrent infection, and 3 (7%) died (2 from nonfunction of the primary graft and 1 from complications of hepatic artery thrombosis). Of all 61 patients given sofosbuvir and ribavirin, 49% had a post-transplantation virologic response. Recurrence was related inversely to the number of consecutive days of undetectable HCV RNA before transplantation. The most frequently reported adverse events were fatigue (in 38% of patients), headache (23%), and anemia (21%). Administration of sofosbuvir and ribavirin before liver transplantation can prevent post-transplant HCV recurrence. ClinicalTrials.gov: NCT01559844.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U1752
Uridine 5′-monophosphate, ≥98%
C9H13N2O9P