EMAIL THIS PAGE TO A FRIEND
Thrombosis research
Microparticle-associated tissue factor activity in patients with acute unprovoked deep vein thrombosis and during the course of one year.
PMID 25262106
Abstract
Tissue factor (TF) is the main in-vivo initiator of blood coagulation. Microparticles (MPs) are small procoagulant membrane vesicles. Elevated TF-bearing MPs have been found in different prothrombotic conditions and MP-associated TF activity may contribute to the pathogenesis of unprovoked deep vein thrombosis (DVT). To determine MP-TF activity levels at diagnosis of DVT and at four additional time points during the course of one year in a well-defined group of patients with unprovoked DVT of the lower limb. In this study, 41 patients with acute unilateral symptomatic and unprovoked DVT of the lower limb were included and followed for 1 year. Venous blood samples for determination of MP-TF activity were drawn at diagnosis of acute DVT, and 1-, 3-, 6-, and 12 months later. In addition, 10 young and healthy control subjects were included. The median MP-TF activity was 0.06 pg/mL (25th-75th percentile: 0.0-0.53) in patients with acute DVT and 0.18 pg/mL (0.07-0.33) in healthy controls, and did not differ significantly (p=0.35). No significant changes in MP-TF activity were found in the follow-up measurements. MP-TF activity did also not differ significantly between patients with proximal- or distal DVT and between those with- or without residual DVT after 6 months. MP-TF activity is low at the acute event in patients with unprovoked DVT of the lower limb and remains unchanged during the course of the disease. Our data do not support the hypothesis that TF-bearing MPs play a determining role in the pathogenesis of unprovoked DVT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3705
Transferrin human, recombinant, expressed in rice