EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Transformations of citrate and Tween coated silver nanoparticles reacted with Na₂S.
PMID 25262296
Abstract
Silver nanoparticles (Ag NPs) are susceptible to transformations in environmental and biological media such as aggregation, oxidation, dissolution, chlorination, sulfidation, formation/replacement of surface coatings following interaction with natural organic matter (NOM). This paper investigates the impact of surface coating and Suwannee River fulvic acid (SRFA) on the transformations and behavior of Ag NPs (citrate coated and Tween coated; cit-Ag NPs and Tween-Ag NPs, respectively), following reaction with different concentrations of Na2S solution (as a source of sulfide species, H2S and HS(-)). These transformations and the dominant mechanisms of transformations were investigated using UV-vis and scanning transmission electron microscopy coupled with electron energy loss spectroscopy. Here, we have shown that Ag NP surface coating impacts their dissolution following dilution in ultrahigh purity water, with higher extent of dissolution of Tween-Ag NPs compared with cit-Ag NPs. Tween-Ag NPs are susceptible to dissolution following their sulfidation at low S/Ag molar ratio. Suwannee River fulvic acid (SRFA) slows down the dissolution of Tween-Ag NPs at low sulfide concentrations and reduces the aggregation of cit-Ag NP in the presence of sodium sulfide. Sulfidation appears to occur by direct interaction of sulfide species with Ag NPs rather than by indirect reaction of sulfide with dissolved Ag species subsequent to dissolution. Furthermore, the sulfidation process results in the formation of partially sulfidized Ag NPs containing unreacted (metallic) subgrains at the edge of the NPs for Tween-Ag NPs in the presence of high sulfide concentration (2000nM Na2S), which occurred to less extent at lower Na2S concentration for Tween-Ag NPs and at all concentrations of Na2S for cit-Ag NPs. Thus, sulfidized Ag NPs may preserve some of the properties of the Ag NPs such as their potential to shed Ag(+) ions and their toxic potential of Ag NPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27109
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
C6H8O7
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
576832
Silver, nanopowder, <100 nm particle size, contains PVP as dispersant, 99.5% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag