EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Modified dextran cross-linked electrospun gelatin nanofibres for biomedical applications.
PMID 25263915
Abstract
Electrospun gelatin nanofibres attract attention of bioengineering arena because of its excellent biocompatibility and structural resemblance with native extracellular matrix. In this study, we have developed gelatin nanofibres using an innovative cross-linking approach to minimize cytotoxic effects. Gelatin was dissolved in water:acetic acid (8:2, v/v) solution and electrospun to form nanofibres with diameter in the range of 156 ± 30 nm. The nanofibres were cross-linked with a modified polysaccharide, namely, dextran aldehyde (DA). Cross-linking with DA could be achieved without compromising the fibrous architecture. DA cross-linked gelatin nanofibres maintained the fibrous morphology in aqueous medium. These mats exhibit improved mechanical properties and gradual degradation behaviour. The nanofibres were evaluated for cytotoxicity, cell adhesion, viability, morphology and proliferation using L-929 fibroblast cells. The results confirmed that DA cross-linked mats were non cytotoxic towards L-929 cells with good cell adhesion, spreading and proliferation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000454
Diacetylfluorescein, European Pharmacopoeia (EP) Reference Standard
C24H16O7
F7378
Fluorescein diacetate, used as cell viability stain
C24H16O7
379921
Hydroxylamine hydrochloride, 99.995% trace metals basis
H3NO · HCl
431362
Hydroxylamine hydrochloride, 99.999% trace metals basis
H3NO · HCl
255580
Hydroxylamine hydrochloride, ACS reagent, 98.0%
H3NO · HCl
159417
Hydroxylamine hydrochloride, ReagentPlus®, 99%
H3NO · HCl
55460
Hydroxylamine hydrochloride, puriss. p.a., ACS reagent, ≥99.0% (RT)
H3NO · HCl
55469
Hydroxylamine hydrochloride, purum p.a., ≥98.0% (RT)
H3NO · HCl
11514
Hydroxylamine hydrochloride, ≥99.9995% (metals basis)
H3NO · HCl
38364
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water
HNa2O4P
P2297
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
C6H3N3O9S
92822
Picrylsulfonic acid solution, 1 M in H2O
C6H3N3O9S
92823
Picrylsulfonic acid solution, ~1 M in H2O
C6H3N3O9S
S1878
Sodium (meta)periodate, ≥99.0%
INaO4
71859
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
INaO4
30323
Sodium (meta)periodate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8%
INaO4
311448
Sodium periodate, ACS reagent, ≥99.8%
INaO4
363642
Sodium periodate, 10 wt. %, matrix silica gel support
INaO4
71639
Sodium phosphate dibasic, BioUltra, anhydrous, ≥99.5% (T)
HNa2O4P
71642
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
HNa2O4P
71496
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
H2NaO4P
S9640
Sodium tetraborate decahydrate, ACS reagent, ≥99.5%
B4Na2O7 · 10H2O
B9876
Sodium tetraborate decahydrate, ReagentPlus®, ≥99.5%
B4Na2O7 · 10H2O
B3545
Sodium tetraborate decahydrate, BioXtra, ≥99.5%
B4Na2O7 · 10H2O
71999
Sodium tetraborate decahydrate, BioUltra, ≥99.5% (T)
B4Na2O7 · 10H2O
31457
Sodium tetraborate decahydrate, puriss. p.a., ACS reagent, reag. ISO, buffer substance, ≥99.5%
B4Na2O7 · 10H2O
11625
Sodium tetraborate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.0-103.0%
B4Na2O7 · 10H2O