EMAIL THIS PAGE TO A FRIEND
Journal of molecular and cellular cardiology
Functional differentiation of RAMP2 and RAMP3 in their regulation of the vascular system.
PMID 25264174
Abstract
Adrenomedullin (AM) is a vasoactive peptide that possesses various bioactivities. AM receptors are dimers consisting of CLR with one of two accessory proteins, RAMP2 or RAMP3. The functional difference between CLR/RAMP2 and CLR/RAMP3 and the relationship between the two receptors remain unclear. To address these issues, we generated RAMP2 and RAMP3 knockout (-/-) mice and have been studying their physiological activities in the vascular system. AM-/- and RAMP2-/- mice die in utero due to blood vessel abnormalities, which is indicative of their essential roles in vascular development. In contrast, RAMP3-/- mice were born normally without any major abnormalities. In adult RAMP3-/- mice, postnatal angiogenesis was normal, but lymphangiography using indocyanine green (ICG) showed delayed drainage of subcutaneous lymphatic vessels. Moreover, chyle transport by intestinal lymphatics was delayed in RAMP3-/- mice, which also showed more severe interstitial edema than wild-type mice in a tail lymphedema model, with characteristic dilatation of lymphatic capillaries and accumulation of inflammatory cells. In scratch-wound assays, migration of isolated RAMP3-/- lymphatic endothelial cells was delayed as compared to wild-type cells, and AM administration failed to enhance the re-endothelialization. The delay in re-endothelialization was due to a primary migration defect rather than a decrease in proliferation. These results suggest that RAMP3 regulates drainage through lymphatic vessels, and that the AM-RAMP3 system could be a novel therapeutic target for controlling postoperative lymphedema.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2633
Cardiogreen, polymethine dye
C43H47N2NaO6S2
21980
Cardiogreen, for microscopy
C43H47N2NaO6S2