PloS one
Reorganisation of the salivary mucin network by dietary components: insights from green tea polyphenols.
PMID 25264771
Abstract
The salivary mucins that include MUC5B (gel-forming) and MUC7 (non-gel-forming) are major contributors to the protective mucus barrier in the oral cavity, and it is possible that dietary components may influence barrier properties. We show how one dietary compound, the green tea polyphenol epigallocatechin gallate (EGCG), can substantially alter the properties of both the polymeric MUC5B network and monomeric MUC7. Using rate-zonal centrifugation, MUC5B in human whole saliva and MUC5B purified from saliva sedimented faster in the presence of EGCG. The faster sedimentation by EGCG was shown to be greater with increasing MUC5B concentration. Particle tracking microrheology was employed to determine the viscosity of purified MUC5B solutions and showed that for MUC5B solutions of 200-1600 µg/mL, EGCG caused a significant increase in mucin viscosity, which was greater at higher MUC5B concentrations. Visualisation of the changes to the MUC5B network by EGCG was performed using atomic force microscopy, which demonstrated increased aggregation of MUC5B in a heterogeneous manner by EGCG. Using trypsin-resistant, high-molecular weight oligosaccharide-rich regions of MUC5B and recombinant N-terminal and C-terminal MUC5B proteins, we showed that EGCG causes aggregation at the protein domains of MUC5B, but not at the oligosaccharide-rich regions of the mucin. We also demonstrated that EGCG caused the majority of MUC7 in human whole saliva to aggregate. Furthermore, purified MUC7 also underwent a large increase in sedimentation rate in the presence of EGCG. In contrast, the green tea polyphenol epicatechin caused no change in the sedimentation rate of either MUC5B or MUC7 in human whole saliva. These findings have demonstrated how the properties of the mucin barrier can be influenced by dietary components. In the case of EGCG, these interactions may alter the function of MUC5B as a lubricant, contributing to the astringency (dry puckering sensation) of green tea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50299
(−)-Epigallocatechin gallate, ≥97.0% (HPLC)
C22H18O11
93894
(−)-Epigallocatechin gallate, analytical standard
C22H18O11
A79604
4-Aminosalicylic acid, 99%
C7H7NO3
746487
Cesium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%
ClCs
203025
Cesium chloride, ≥99.999% trace metals basis
ClCs
449733
Cesium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClCs
289329
Cesium chloride, ReagentPlus®, 99.9%
ClCs
C3139
Cesium chloride, optical grade, ≥99.5% trace metals basis
ClCs
C3011
Cesium chloride, Grade I, ≥99.0%
ClCs
562599
Cesium chloride, 99.99% trace metals basis
ClCs
562602
Cesium chloride, Cabot high-purity grade
ClCs
C3032
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
ClCs
C4036
Cesium chloride, for molecular biology, ≥98%
ClCs
C3309
Cesium chloride, BioXtra, ≥99.5% (titration)
ClCs
C6914
Cesium chloride, Grade II, ≥98%
ClCs
20966
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClCs
20967
Cesium chloride, BioUltra, ≥99.0% (AT)
ClCs
31807
Cesium chloride, puriss. p.a., ≥99.5%
ClCs
03970590
Epigallocatechin gallate, primary pharmaceutical reference standard
C22H18O11
PHR1333
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
C22H18O11