EMAIL THIS PAGE TO A FRIEND
Chinese medical sciences journal = Chung-kuo i hsueh k'o hsueh tsa chih
ZM-66, a new podophyllotoxin derivative inhibits proliferation and induces apoptosis in K562/ADM cells.
PMID 25264886
Abstract
To investigate the anti-tumor effect of ZM-66 on multidrug-resistant leukemic cell line K562/ADM. The K562/ADM cells were treated with varying concentrations (0, 1, 2, 4 × 10⁻³ mmol/L) of ZM-66 or etoposide for 24 hours. The proliferation was detected by Sulforhodamine B Sodium Salt (SRB) assay and apoptosis was detected by flow cytometry analysis and fluorescent staining. In addition, the expression levels of p53 and bax genes in K562/ADM cells were detected by RT-PCR analysis. The level of P-glycoprotein (P-gp), P53 and Bax protein in K562/ADM cells were detected by Western blot assay. SRB assay demonstrated that etoposide had little inhibitory effect on K562/ADM cells, whereas ZM-66 (1, 2, 4 × 10⁻³ mmol/L) had significantly inhibitory effect on K562/ADM cells (all P<0.01). The acridine orange/propidium iodide dual staining showed that there were typical condensation of chromatin and nuclear fragmentation nuclei with red color in ZM-66 treated cells. Flow cytometric analysis showed that there was a significantly increase of apoptotic cells in K562/ADM cells after treated with ZM-66. RT-PCR showed that the p53 and bax mRNA expression levels in K562/ADM cells treated with ZM-66 at 1, 2, 4 × 10⁻³ mmol/L were higher than those in the cell without treatment. Western blot showed that the P53 and Bax protein expression levels in K562/ADM cells treated with ZM-66 at 2, 4 × 10⁻³ mmol/L were higher than those in the cell without treatment. But the P-gp protein expression level in K562/ADM cells treated with ZM-66 at 2, 4 × 10⁻³ mmol/L was gradually lower than those in the cell without treatment. ZM-66 is able to induce cell death by apoptosis in vitro, as a result of the reverse of the apoptosis resistance in drug-resistant K562/ADM cells by modulating expression of key factors associated with apoptosis induction.