PloS one
MiR-122 directly inhibits human papillomavirus E6 gene and enhances interferon signaling through blocking suppressor of cytokine signaling 1 in SiHa cells.
PMID 25265013
Abstract
Human Papillomavirus (HPV) 16 infection is considered as one of the significant causes of human cervical cancer. The expression of the viral oncogenes like E6 and E7 play an important role in the development of the cancer. MiR-122 has been reported to exhibit a strong relationship with hepatitis viruses and take part in several tumor development, while the effects of miR-122 on HPV infection and the HPV viral oncogenes expression still remain unexplored. In this study, using RNAhybrid software, the potential binding sites between miR-122 and HPV16 E6 and E7 mRNAs were identified. Over and loss of miR-122 function showed that miR-122 could directly bind with HPV16 E6 mRNA and significantly inhibit its expression in SiHa cells, which was further confirmed by constructing the miR-122-E6-mu to eliminate the miR-122 binding effects with E6. The increase of the expression of type I interferon (IFN) and its classical effective molecules and the phosphorylation of signal transducers and activators of transcription (STAT1) protein indicated that miR-122 might enhance type I interferon in cervical carcinoma cells, which explained the significant reduction of HPV16 E7 and E6*I mRNA expression. This might be due to the binding between miR-122 and suppressor of cytokine signaling 1 (SOCS1) mRNA, which is the suppressor of interferon signaling pathway. Moreover, it was identified that the miR-122 binding position was nt359-nt375 in SOCS1 mRNA. Taken together, this study indicated that HPV16 could be effectively inhibited by miR-122 through both direct binding with E6 mRNA and promoting SOCS1-dependent IFN signaling pathway. Thus, miR-122 may serve as a new therapeutic option for inhibiting HPV infection.