PloS one
Small dense low density lipoprotein particles are associated with poor outcome after angioplasty in peripheral artery disease.
PMID 25265512
Abstract
In patients suffering from symptomatic peripheral artery disease (PAD), percutaneous revascularization is the treatment of choice. However, restenosis may occur in 10 to 60% in the first year depending on a variety of factors. Small dense low density lipoprotein (sdLDL) particles are associated with an increased risk for cardiovascular events, but their role in the process of restenosis is not known. We conducted a prospective study to analyze the association of sdLDL particles with the outcome of balloon angioplasty in PAD. The composite primary endpoint was defined as improved walking distance and absence of restenosis. Patients with angiographically documented PAD of the lower extremities who were scheduled for lower limb revascularization were consecutively recruited for the study. At baseline and at three month follow-up triglyceride, total cholesterol, LDL size and subclasses and HDL cholesterol and ankle-brachial index (ABI) were measured. Three months after the intervention duplex sonography was performed to detect restenosis. Sixty-four patients (53% male) with a mean age of 68.6±9.9 years were included. The proportion of small- dense LDL particles (class III and IV) was significantly lower (33.1±11.0% vs. 39.4±12.1%, p = 0.038) in patients who reached the primary end-point compared with those who did not. Patients with improved walking distance and without restenosis had a significantly higher LDL size at baseline (26.6±1.1 nm vs. 26.1±1.1 nm, p = 0.046) and at follow-up (26.7±1.1 nm vs. 26.2±0.9 nm, p = 0.044) than patients without improvement. Small-dense LDL particles are associated with worse early outcome in patients undergoing percutaneous revascularization for symptomatic PAD.