EMAIL THIS PAGE TO A FRIEND
BMC immunology
T lymphocytes maintain structure and function of fibroblastic reticular cells via lymphotoxin (LT)-B.
PMID 25266629
Abstract
Although a lot is known about how Fibroblastic Reticular Cells (FRCs) can regulate T lymphocytes (T cells), little is understood about whether or how T cells can regulate FRCs. This study shows that the absence of T cells inhibited the secretion of ER-TR7 by splenic FRCs, induced the structural disorder of FRCs, down-regulated the expression of the chemokine ligands CCL21 and CCL19, and weakened the homing ability of T cells to the spleen of nude mice. Transfusion of T cells from BALB/c mice restored the structure and functions of FRCs and recovered them. The expression of lymphotoxin (LT)-B was significantly downregulated in the absence of T cells from nude mice and was recovered after the transfusion of T cells. After the occlusion of the LT-B receptor, the FRCs' structure and functions were not restored by transfusion of T cells. These data reveal that the absence of T cells will subject spleen FRCs to structural and functional abnormality, and weaken the homing ability of T cells to the spleen. These changes are attributed to the T-cell- derived LT-B.