EMAIL THIS PAGE TO A FRIEND
Human reproduction (Oxford, England)
Cell-free DNA in human follicular fluid as a biomarker of embryo quality.
PMID 25267787
Abstract
Could cell-free DNA (cfDNA) quantification in individual human follicular fluid (FF) samples become a new non-invasive predictive biomarker for in vitro fertilization (IVF) outcomes? CfDNA level in human follicular fluid samples was significantly correlated with embryo quality and could be used as an innovative non-invasive biomarker to improve IVF outcomes. CfDNA fragments, resulting from apoptotic or necrotic events, are present in the bloodstream and their quantification is already used as a biomarker for gynaecological and pregnancy disorders. Follicular fluid is important for oocyte development and contains plasma components and factors secreted by granulosa cells during folliculogenesis. CfDNA presence in follicular fluid and its potential use as an IVF outcome biomarker have never been investigated. One hundred individual follicular fluid samples were collected from 43 female patients undergoing conventional IVF (n = 26) or ICSI (n = 17). CfDNA level was quantified in each individual follicular fluid sample. At oocyte collection day, follicles were aspirated individually. Only blood-free follicular fluid samples were included in the study. Follicle size was calculated based on the follicular fluid volume. Each corresponding cumulus-oocyte complex was isolated for IVF or ICSI procedures. Follicular fluid cfDNA was measured by quantitative PCR with ALU-specific primers. Human follicular fluid samples from individual follicles contain measurable amounts of cfDNA (mean ± SD, 1.62 ± 2.08 ng/µl). CfDNA level was significantly higher in small follicles (8-12 mm in diameter) than in large ones (>18 mm) (mean ± SD, 2.54 ± 0.78 ng/µl versus 0.71 ± 0.44 ng/µl, respectively, P = 0.007). Moreover, cfDNA concentration was significantly and negatively correlated with follicle size (r = -0.34; P = 0.003). A weak significant negative correlation between DNA integrity and 17β-estradiol level in follicular fluid samples at oocyte collection day was observed (r = -0.26; P = 0.008). CfDNA level in follicular fluid samples corresponding to top quality embryos was significantly lower than in follicular fluid samples related to poor quality embryos (P = 0.022). Similarly, cfDNA level was also significantly lower in follicular fluid samples related to embryos with low fragmentation rate (≤25%) than with high fragmentation rate (>25%) (P = 0.02). A larger study should be conducted in order to establish the predictive value of cfDNA level for embryo quality and to investigate whether follicular fluid cfDNA levels are correlated with embryo implantation rates and pregnancy outcomes. Moreover, the role of follicular fluid cfDNA on embryo quality should be studied to determine whether high cfDNA concentration in follicular fluid is only a consequence or also a cause of follicular dysfunction. CfDNA evaluation in individual follicular fluid samples might represent an innovative biomarker of embryo quality to use as a supplemental tool to predict embryo quality during IVF. This study was partially supported by the University Hospital of Montpellier and Ferring Pharmaceuticals. The authors of the study have no competing interests to report.