EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Neutrophils counteract autophagy-mediated anti-inflammatory mechanisms in alveolar macrophage: role in posthemorrhagic shock acute lung inflammation.
PMID 25267975
Abstract
Acute lung injury (ALI) is a major component of multiple organ dysfunction syndrome after hemorrhagic shock (HS) resulting from major surgery and trauma. The increased susceptibility in HS patients to the development of ALI suggests not yet fully elucidated mechanisms that enhance proinflammatory responses and/or suppress anti-inflammatory responses in the lung. Alveolar macrophages (AMϕ) are at the center of the pathogenesis of ALI after HS. We have previously reported that HS-activated polymorphonuclear neutrophils (PMNs) interact with macrophages to influence inflammation progress. In this study, we explore a novel function of PMNs regulating AMϕ anti-inflammatory mechanisms involving autophagy. Using a mouse "two-hit" model of HS/resuscitation followed by intratracheal injection of muramyl dipeptide, we demonstrate that HS initiates high mobility group box 1/TLR4 signaling, which upregulates NOD2 expression in AMϕ and sensitizes them to subsequent NOD2 ligand muramyl dipeptide to augment lung inflammation. In addition, upregulated NOD2 signaling induces autophagy in AMϕ, which negatively regulates lung inflammation through feedback suppression of NOD2-RIP2 signaling and inflammasome activation. Importantly, we further demonstrate that HS-activated PMNs that migrate in alveoli counteract the anti-inflammatory effect of autophagy in AMϕ, possibly through NAD(P)H oxidase-mediated signaling to enhance I-κB kinase γ phosphorylation, NF-κB activation, and nucleotide-binding oligomerization domain protein 3 inflammasome activation, and therefore augment post-HS lung inflammation. These findings explore a previously unidentified complexity in the mechanisms of ALI, which involves cell-cell interaction and receptor cross talk.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000440
Anti-CD45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405577
Anti-CD8A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037756
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408221
Anti-CIAS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA012878
Anti-NLRP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA017374
Anti-NLRP3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410191
Anti-NLRP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21465 Anti-PTPRC antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4700471 Monoclonal Anti-CD45 antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution
AMAB90518
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0159, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90519
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0160, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700587 Monoclonal Anti-CD45 antibody produced in rat, clone EM-05, purified immunoglobulin, buffered aqueous solution
C7423
Monoclonal Anti-CD8 antibody produced in mouse, clone UCHT-4, purified immunoglobulin, buffered aqueous solution
SAB4700084 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-31, purified immunoglobulin, buffered aqueous solution
SAB4700089 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-87, purified immunoglobulin, buffered aqueous solution
SAB1402142
Monoclonal Anti-CD8A antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
SAB1402652
Monoclonal Anti-CIAS1, (N-terminal) antibody produced in mouse, clone 3B1, purified immunoglobulin, buffered aqueous solution
SAB4700649 Monoclonal Anti-PTPRC antibody produced in mouse, clone IVA103, purified immunoglobulin, buffered aqueous solution