EMAIL THIS PAGE TO A FRIEND
Ultrastructural pathology
Brain metastasis of crystal-deficient, CD68-positive alveolar soft part sarcoma: ultrastructural features and differential diagnosis.
PMID 25268941
Abstract
We report a case of alveolar soft part sarcoma (ASPS) presenting as an isolated frontal lobe metastasis. The tumor demonstrated little or no immunoreactivity for a broad panel of antibodies yet strong, diffuse immunoreactivity with CD68. On electron microscopy, the characteristic rectangular to rhomboid crystalline inclusions of ASPS were not present. Electron-dense granules resembling peroxisomes were present, sometimes in association with elongated granular structures having a periodic, lattice-like arrangement. Metastatic ASPS was confirmed by demonstration of an ASPSCR1-TFE3 fusion and imaging studies that excluded metastatic Xp11.2 translocation renal cell carcinoma. The primary site was subsequently identified in the lower extremity.