EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Microfluidic Western blotting of low-molecular-mass proteins.
PMID 25268977
Abstract
We describe a microfluidic Western blot assay (μWestern) using a Tris tricine discontinuous buffer system suitable for analyses of a wide molecular mass range (6.5-116 kDa). The Tris tricine μWestern is completed in an enclosed, straight glass microfluidic channel housing a photopatterned polyacrylamide gel that incorporates a photoactive benzophenone methacrylamide monomer. Upon brief ultraviolet (UV) light exposure, the hydrogel toggles from molecular sieving for size-based separation to a covalent immobilization scaffold for in situ antibody probing. Electrophoresis controls all assay stages, affording purely electronic operation with no pumps or valves needed for fluid control. Electrophoretic introduction of antibody into and along the molecular sieving gel requires that the probe must traverse through (i) a discontinuous gel interface central to the transient isotachophoresis used to achieve high-performance separations and (ii) the full axial length of the separation gel. In-channel antibody probing of small molecular mass species is especially challenging, since the gel must effectively sieve small proteins while permitting effective probing with large-molecular-mass antibodies. To create a well-controlled gel interface, we introduce a fabrication method that relies on a hydrostatic pressure mismatch between the buffer and polymer precursor solution to eliminate the interfacial pore-size control issues that arise when a polymerizing polymer abuts a nonpolymerizing polymer solution. Combined with a new swept antibody probe plug delivery scheme, the Tris tricine μWestern blot enables 40% higher separation resolution as compared to a Tris glycine system, destacking of proteins down to 6.5 kDa, and a 100-fold better signal-to-noise ratio (SNR) for small pore gels, expanding the range of applicable biological targets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3699
Acrylamide/Bis-acrylamide, 30% solution, BioReagent, suitable for electrophoresis, 37.5:1
SAB1300902
Anti-APS (center) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1405474
Anti-KLK3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000764
Anti-KLK3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410703
Anti-KLK3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4501649
Anti-KLKB1 (heavy chain, Cleaved-Arg390) antibody produced in rabbit, affinity isolated antibody
HPA005634
Anti-KLKB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV45678
Anti-KLKB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2106715
Anti-KLKB1 antibody produced in rabbit, affinity isolated antibody
SAB4501531
Anti-Prostate-specific Antigen antibody produced in rabbit, affinity isolated antibody
WH0000354M2 Monoclonal Anti-KLK3 antibody produced in mouse, clone 1B1, purified immunoglobulin, buffered aqueous solution
SAB5300268
Monoclonal Anti-KLK3 antibody produced in mouse, clone 5A11E9, ascites fluid
294381
N,N′-(1,2-Dihydroxyethylene)bisacrylamide, 97%
C8H12N2O4
T1165 Tris-Tricine-SDS Buffer 10× Concentrate