EMAIL THIS PAGE TO A FRIEND
Current drug delivery
Self-adjuvanting therapeutic peptide-based vaccine induce CD8+ cytotoxic T lymphocyte responses in a murine human papillomavirus tumor model.
PMID 25269453
Abstract
Vaccine candidates for the treatment of human papillomavirus (HPV)-associated cancers are aimed to activate T-cells and induce development of cytotoxic anti-tumor specific responses. Peptide epitopes derived from HPV-16 E7 oncogenic protein have been identified as promising antigens for vaccine development. However, peptide-based antigens alone elicit poor cytotoxic T lymphocyte (CTL) responses and need to be formulated with an adjuvant (immunostimulant) to achieve the desired immune responses. We have reported the ability of polyacrylate 4-arm star-polymer (S4) conjugated with HPV-16 E744-57 (8Qmin) epitope to reduce and eradicate TC-1 tumor in the mouse model. Herein, we have studied the mechanism of induction of immune responses by this polymer-peptide conjugate and found prompt uptake of conjugate by antigen presenting cells, stimulating stronger CD8(+) rather than CD4(+) or NK cell responses.