EMAIL THIS PAGE TO A FRIEND
Cancer research
MPHOSPH1: a potential therapeutic target for hepatocellular carcinoma.
PMID 25269478
Abstract
MPHOSPH1 is a critical kinesin protein that functions in cytokinesis. Here, we show that MPHOSPH1 is overexpressed in hepatocellular carcinoma (HCC) cells, where it is essential for proliferation. Attenuating MPHOSPH1 expression with a tumor-selective shRNA-expressing adenovirus (Ad-shMPP1) was sufficient to arrest HCC cell proliferation in a manner associated with an accumulation of multinucleated polyploid cells, induction of postmitotic apoptosis, and increased sensitivity to taxol cytotoxicity. Mechanistic investigations showed that attenuation of MPHOSPH1 stabilized p53, blocked STAT3 phosphorylation, and prolonged mitotic arrest. In a mouse subcutaneous xenograft model of HCC, tumoral injection of Ad-shMPP1 inhibited MPHOSPH1 expression and tumor growth in a manner correlated with induction of apoptosis. Combining Ad-shMPP1 injection with taxol administration enhanced antitumor efficacy relative to taxol alone. Furthermore, Ad-shMPP1 tail vein injection suppressed formation of orthotopic liver nodules and prevented hepatic dysfunction. Taken together, our results identify MPHOSPH1 as an oncogenic driver and candidate therapeutic target in HCC.