EMAIL THIS PAGE TO A FRIEND
Transplant international : official journal of the European Society for Organ Transplantation
Effectiveness of porcine dermal collagen in giant hernia closure in patients with deleterious fascia constitution after orthotopic liver transplantation.
PMID 25269850
Abstract
Incisional hernias (IHs) occur universally after orthotopic liver transplantation (OLT). This study aimed to investigate the effectiveness of porcine dermal collagen (PDC) as a closing aid in giant hernias after OLT in a prospective trial. If direct closure (DC) was not feasible due to the hernia size and abdominal wall constitution, a PDC mesh was implanted. All patients from the PDC and DC groups were followed prospectively for 24xa0months. IH recurrence rates served as the primary endpoint, and the development of infections and wound healing disorders served as the secondary endpoints. Recurrence rate was 21% (4/19) in DC patients and 12% (2/16) in PDC patients (Pxa0=xa00.045). Implant site infections occurred in five of PDC and one of DC patients (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34218
Cefuroxime, VETRANAL, analytical standard
C16H16N4O8S