EMAIL THIS PAGE TO A FRIEND
The International journal of neuroscience
The Nitric oxide/CGMP/KATP pathway mediates systemic and central antinociception induced by resistance exercise in rats.
PMID 25271801
Abstract
Resistance exercise (RE) is characterized to increase strength, tone, mass, and/or muscular endurance and also for produces many beneficial effects, such as blood pressure and osteoporosis reduction, diabetes mellitus control, and analgesia. However, few studies have investigated endogenous mechanisms involved in the RE-induced analgesia. Thus, the aim of this study was evaluate the role of the NO/CGMP/KATP pathway in the antinociception induced by RE. Wistar rats were submitted to acute RE in a weight-lifting model. The nociceptive threshold was measured by mechanical nociceptive test (paw-withdrawal). To investigate the involvement of the NO/CGMP/KATP pathway the following nitric oxide synthase (NOS) non-specific and specific inhibitors were used: N-nitro-l-arginine (NOArg), Aminoguanidine, N5-(1-Iminoethyl)-l-ornithine dihydrocloride (l-NIO), Nω-Propyl-l-arginine (l-NPA); guanylyl cyclase inhibitor, 1H-[1,2,4]oxidiazolo[4,3-a]quinoxalin-1-one (ODQ); and KATP channel blocker, Glybenclamide; all administered subcutaneously, intrathecally and intracerebroventricularly. Plasma and cerebrospinal fluid (CSF) nitrite levels were determined by spectrophotometry. The RE protocol produced antinociception, which was significantly reversed by NOS specific and unspecific inhibitors, guanylyl cyclase inhibitor (ODQ) and KATP channel blocker (Glybenclamide). RE was also responsible for increasing nitrite levels in both plasma and CSF. These finding suggest that the NO/CGMP/KATP pathway participates in antinociception induced by RE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0639
Glybenclamide, ≥99% (HPLC)
C23H28ClN3O5S
G2539
Glyburide, meets USP testing specifications
C23H28ClN3O5S