EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Characterization of ritonavir-mediated inactivation of cytochrome P450 3A4.
PMID 25274602
Abstract
Ritonavir is a human immunodeficiency virus (HIV) protease inhibitor and an inhibitor of cytochrome P450 3A4, the major human hepatic drug-metabolizing enzyme. Given the potent inhibition of CYP3A4 by ritonavir, subtherapeutic doses of ritonavir are used to increase plasma concentrations of other HIV drugs oxidized by CYP3A4, thereby extending their clinical efficacy. However, the mechanism of inhibition of CYP3A4 by ritonavir remains unclear. To date, data suggests multiple types of inhibition by ritonavir, including mechanism-based inactivation by metabolic-intermediate complex formation, competitive inhibition, irreversible type II coordination to the heme iron, and more recently heme destruction. The results presented here demonstrate that inhibition of CYP3A4 by ritonavir occurs by CYP3A4-mediated activation and subsequent formation of a covalent bond to the apoprotein. Incubations of [(3)H]ritonavir with reconstituted CYP3A4 and human liver microsomes resulted in a covalent binding stoichiometry equal to 0.93 ± 0.04 moles of ritonavir bound per mole of inactivated CYP3A4. The metabolism of [(3)H]ritonavir by CYP3A4 leads to the formation of a covalent adduct specifically to CYP3A4, confirmed by radiometric liquid chromatography-trace and whole-protein mass spectrometry. Tryptic digestion of the CYP3A4-[(3)H]ritonavir incubations exhibited an adducted peptide (255-RM K: ESRLEDTQKHR-268) associated with a radiochromatic peak and a mass consistent with ritonavir plus 16 Da, in agreement with the whole-protein mass spectrometry. Additionally, nucleophilic trapping agents and scavengers of free oxygen species did not prevent inactivation of CYP3A4 by ritonavir. In conclusion, ritonavir exhibited potent time-dependent inactivation of CYP3A, with the mechanism of inactivation occurring though a covalent bond to Lys257 of the CYP3A4 apoprotein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0491
Ritonavir, ≥98% (HPLC)
C37H48N6O5S2
Y0000714 Ritonavir, European Pharmacopoeia (EP) Reference Standard
C37H48N6O5S2