EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Emerging ceramic-based materials for dentistry.
PMID 25274751
Abstract
Our goal is to give an overview of a selection of emerging ceramics and issues for dental or biomedical applications, with emphasis on specific challenges associated with full-contour zirconia ceramics, and a brief synopsis on new machinable glass-ceramics and ceramic-based interpenetrating phase composites. Selected fabrication techniques relevant to dental or biomedical applications such as microwave sintering, spark plasma sintering, and additive manufacturing are also reviewed. Where appropriate, the authors have added their opinions and guidance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
419141
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zr
403296
Zirconium, powder, −100 mesh
Zr
267724
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zr
267651
Zirconium, sponge, ≥99% trace metals basis
Zr
14602
Zirconium, ≥97.0% Zr + Hf basis, powder
Zr
756385
Zirconium, powder, in water, -325 mesh, 99.5% trace metals basis (excluding Hf)
Zr
GF69451487
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zr
GF27504402
Zirconium, foil, 300x300mm, thickness 0.5mm, annealed, 99.2%
Zr
GF74069837
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zr
GF13629483
Zirconium, foil, 50x50mm, thickness 0.1mm, annealed, 99.2%
Zr
GF87053474
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zr
GF83603909
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zr
GF04051886
Zirconium, foil, 50x50mm, thickness 2.0mm, annealed, 99.2%
Zr
GF14815593
Zirconium, foil, 10mm disks, thickness 0.0125mm, 99.2%
Zr
GF58113019
Zirconium, foil, 10mm disks, thickness 0.015mm, 99.2%
Zr
GF67870468
Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%
Zr
GF58442070
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF54823642
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF75562869
Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%
Zr
GF55287221
Zirconium, foil, 10mm disks, thickness 0.02mm, 99.2%
Zr
GF38009044
Zirconium, foil, 10mm disks, thickness 0.05mm, annealed, 99.2%
Zr
GF89139594
Zirconium, foil, 10mm disks, thickness 0.075mm, annealed, 99.2%
Zr
GF22007595
Zirconium, foil, 10mm disks, thickness 0.125mm, annealed, 99.2%
Zr
GF50038039
Zirconium, foil, 10mm disks, thickness 0.25mm, annealed, 99.2%
Zr
GF03886443
Zirconium, foil, 10mm disks, thickness 0.5mm, annealed, 99.2%
Zr
GF09475027
Zirconium, foil, 15mm disks, thickness 0.009mm, 99.2%
Zr
GF51948872
Zirconium, foil, 15mm disks, thickness 0.0125mm, 99.2%
Zr
GF83808221
Zirconium, foil, 15mm disks, thickness 0.016mm, 99.2%
Zr
GF05259135
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zr
GF72812370
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zr