EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Pro-brain-derived neurotrophic factor inhibits GABAergic neurotransmission by activating endocytosis and repression of GABAA receptors.
PMID 25274828
Abstract
GABA is the canonical inhibitory neurotransmitter in the CNS. This inhibitory action is largely mediated by GABA type A receptors (GABAARs). Among the many factors controlling GABAergic transmission, brain-derived neurotrophic factor (BDNF) appears to play a major role in regulating synaptic inhibition. Recent findings have demonstrated that BDNF can be released as a precursor (proBDNF). Although the role of mature BDNF on GABAergic synaptogenesis and maintenance has been well studied, an important question still unanswered is whether secreted proBDNF might affect GABAergic neurotransmission. Here, we have used 14 d in vitro primary culture of hippocampal neurons and ex vivo preparations from rats to study the function of proBDNF in regulation of GABAAR trafficking and activity. We demonstrate that proBDNF impairs GABAergic transmission by the activation of two distinct pathways: (1) a RhoA-Rock-PTEN pathway that decreases the phosphorylation levels of GABAAR, thus affecting receptor function and triggering endocytosis and degradation of internalized receptors, and (2) a JAK-STAT-ICER pathway leading to the repression of GABAARs synthesis. These effects lead to the diminution of GABAergic synapses and are correlated with a decrease in GABAergic synaptic currents. These results revealed new functions for proBDNF-p75 neurotrophin receptor signaling pathway in the control of the efficacy of GABAergic synaptic activity by regulating the trafficking and synthesis of GABAARs at inhibitory synapses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100649
Anti-EEA1 antibody produced in rabbit, affinity isolated antibody
SAB4300682
Anti-EEA1 antibody produced in rabbit, affinity isolated antibody
HPA038158
Anti-EEA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA038159
Anti-EEA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500694
Anti-NGFR antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2502011
Anti-NGFR antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4503908
Anti-phospho-CREB (pSer121) antibody produced in rabbit, affinity isolated antibody
SAB4504375
Anti-phospho-CREB (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB4503757
Anti-phospho-CREB (pSer142) antibody produced in rabbit, affinity isolated antibody
SAB4504577
Anti-phospho-CREB (pThr100) antibody produced in rabbit, affinity isolated antibody
SAB1306301 ANTI-PHOSPHO-CREB(S133) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300217
Anti-phospho-CREB1 (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4300040
Anti-phospho-CREB1 (pSer133) antibody produced in rabbit, affinity isolated antibody
SAB2107401
Anti-VAMP1 antibody produced in rabbit, affinity isolated antibody
SAB2107402
Anti-VAMP1 antibody produced in rabbit, affinity isolated antibody
SRP3014 BDNF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
B3795 Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
WH0001385M1
Monoclonal Anti-CREB1 antibody produced in mouse, clone 3F1-1B2, purified immunoglobulin, buffered aqueous solution
SAB1403692
Monoclonal Anti-CREB1, (N-terminal) antibody produced in mouse, clone 5F2, purified immunoglobulin, buffered aqueous solution
N0750000
Nifedipine, European Pharmacopoeia (EP) Reference Standard
C17H18N2O6
1463508
Nifedipine, United States Pharmacopeia (USP) Reference Standard
C17H18N2O6
PHR1290
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
C17H18N2O6
SRP6053 ProBDNF human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)