EMAIL THIS PAGE TO A FRIEND
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society
Analyzing simultaneous positive expression of EZH2 and P53 protein to improve predictive value in cervical squamous cell carcinoma.
PMID 25275664
Abstract
The current study was undertaken to investigate the predictive value of simultaneous enhancer of zeste homolog 2 (EZH2) and P53 expression in lesions of patients with cervical squamous cell carcinoma. Quantum dot double fluorescence staining was applied to detect EZH2 and P53 protein in biopsy tissue of 168 patients with cervical squamous cell carcinoma. The expression was classified into double positive (EZH2 and P53 were positively expressed), single positive (either EZH2 or P53 was positively expressed), and double negative (neither was positively expressed). The EZH2 and P53 expression, clinical stages of cervical cancer, lymph node metastasis, progression-free survival, and overall survival were analyzed. A relationship of EZH2 and P53 expression with the clinical stage of cervical cancer and lymph node metastasis was indicated. Simultaneous detection of both proteins could partly predict prognosis, consistent with previous studies' results. The current study demonstrated that the expression levels of EZH2 and P53 in tumor tissue and the proportion of cases with double-positive expression significantly increased with increasing clinical stages of cancer, also confirming up-regulated expression of EZH2 and P53 with increasing stages. These findings may suggest that the 2 proteins were involved in the development of cervical cancer, but the nature of their interaction is undefined. The increase of EZH2 and P53 expression in patients with lymph node metastasis indicated that they may be involved in metastasis of cervical cancer. Simultaneous positive EZH2 and P53 expression could improve the predictive value of a poor prognosis in cervical cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7031
Anti-EZH2 (N-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405776
Anti-EZH2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
E6906
Anti-EZH2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV38470
Anti-EZH2 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB2106436
Anti-EZH2 antibody produced in rabbit, affinity isolated antibody
SAB1410354
Anti-EZH2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
SAB4300363
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2102511
Anti-TP53 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102512
Anti-TP53 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2102513
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2102514
Anti-TP53 (ab4) antibody produced in rabbit, affinity isolated antibody
SAB2102515
Anti-TP53 (ab5) antibody produced in rabbit, affinity isolated antibody
SAB1406544
Anti-TP53 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV02055
Anti-TP53 antibody produced in rabbit, IgG fraction of antiserum
AV37163
Anti-TRP53 antibody produced in rabbit, IgG fraction of antiserum
SAB2106418
Anti-TRP53 antibody produced in rabbit, affinity isolated antibody
SAB1408985 Monoclonal Anti-EZH2 antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
AMAB90956
Monoclonal Anti-p53 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2199, purified immunoglobulin, buffered aqueous glycerol solution
WH0007157M1
Monoclonal Anti-TP53 antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1404483
Monoclonal Anti-TP53 antibody produced in mouse, clone 2C11, purified immunoglobulin, buffered aqueous solution