EMAIL THIS PAGE TO A FRIEND
SpringerPlus
Coating of magnetite with mercapto modified rice hull ash silica in a one-pot process.
PMID 25279307
Abstract
In this research, mercapto-silica coated magnetite (Fe3O4-SiO2-SH) has been prepared in aqueous solution through a simple approach so called a one-pot process. The Fe3O4-SiO2-SH was prepared in nitrogen condition by mixing magnetite, 3-mercaptopropyltrimethoxysilane (MPTMS), and sodium silicate (Na2SiO3) solution extracted from rice hull ash, and adjusting the pH of 7.0 using hydrochloric acid. The residue was washed with deionized water, dried at 150°C and separated with an external magnetic field. In that work, the volume of MPTMS and Na2SiO3 was varied and the total amount of Si represented as silica was kept constant. Characters of the material including the functional group presence, the structure, the porosity, the morphology and stability toward various solvents were identified and evaluated. Results of characterization indicated that mercapto-silica has been coated magnetite particle with a simple one-pot process. Coating mercapto-silica on magnetite increases particle size, surface area, and chemical stability. Additionally, Fe3O4-SiO2-SH also shows high stability toward various organic solvents. The magnetic property of magnetite does not change after coating and the addition of nonmagnetic material still gives high value of maximum saturation magnetization. The presence of mercapto groups effective for interaction with heavy metal ions, the high chemical stability without removing the magnetic property promises the prospective application of Fe3O4-SiO2-SH in the future such as for separation and removal of heavy metal ions from aquatic environments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175617
(3-Mercaptopropyl)trimethoxysilane, 95%
C6H16O3SSi
700312
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700320
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700304
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
725331
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725358
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725366
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
747327
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747424
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm diameter, biotin functionalized, 1 mg/mL Fe, dispersion in H2O
Fe3O4
747254
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion
Fe3O4
747343
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747319
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747300
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747408
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), PEG functionalized, dispersion, 1 mg/mL Fe in H2O
Fe3O4
747416
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), biotin functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
790508
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
518158
Iron(II,III) oxide, 99.99% trace metals basis
Fe3O4
637106
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Fe3O4
310069
Iron(II,III) oxide, powder, <5 μm, 95%
Fe3O4