EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Formation of trichloronitromethane and dichloroacetonitrile in natural waters: precursor characterization, kinetics and interpretation.
PMID 25279758
Abstract
During the chloramination of natural waters, both chloramines and dissolved organic nitrogen (DON) can serve as nitrogen sources for the formation of trichloronitromethane (TCNM) and dichloroacetonitrile (DCAN). The present study investigated the formation kinetics and precursor characteristics of TCNM and DCAN. (15)N-Isotopic monochloramination of the organic fractions produced both (15)N- and (14)N-DCAN and TCNM. Nitrogenous disinfection byproduct (N-DBP) formation, in which the nitrogen precursor originated from DON ((14)N-DCAN and (14)N-TCNM), followed a second-order reaction kinetics (k=3.2×10(-5) to 9.4×10(-5)μM(-1)h(-1)). The formation of N-DBP where the nitrogen atoms originated from chloramines (e.g. (15)N-DCAN and (15)N-TCNM) correlated linearly with chloramine exposure. The discrepancy in formation kinetics results in that the (14)N-DCAN concentrations were two to ten times higher than (15)N-DCAN in the beginning of the reaction (<12h). Possible rate equations are proposed in this study. The results of a model compound study support the results of the chloramination of natural waters. In addition, 4-hydroxybenzaldehyde, an oxidative product commonly found during chlorination/chloramination of natural organic matters, gave a 10-fold greater yield of DCAN than that produced from tyrosine; 4-hydroxybenzaldehyde is thus an important precursor in DCAN formation by chloramine incorporation during the chloramination of natural waters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C94009
4-Cyanophenol, 95%
C7H5NO
144088
4-Hydroxybenzaldehyde, 98%
C7H6O2
54590
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
C7H6O2
W398403
4-Hydroxybenzaldehyde, ≥97%, FG
C7H6O2
91554
4-Hydroxybenzaldehyde, analytical standard
C7H6O2
45528
4-Hydroxybenzonitrile, PESTANAL®, analytical standard
C7H5NO
H21101
4-Hydroxyphenylacetonitrile, 98%
C8H7NO
22754
Chlorine, puriss., ≥99.8%
Cl2
295132
Chlorine, ≥99.5%
Cl2
T3754
L-Tyrosine, reagent grade, ≥98% (HPLC)
C9H11NO3
T8566
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, ≥99.0%
C9H11NO3
T4321
L-Tyrosine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C9H11NO3
93829
L-Tyrosine, BioUltra, ≥99.0% (NT)
C9H11NO3
W373605
L-Tyrosine, ≥97%, FG
C9H11NO3
91515
L-Tyrosine, certified reference material, TraceCERT®
C9H11NO3
PHR1097
L-Tyrosine
C9H11NO3
1705006
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
C9H11NO3
261513
N,N-Diethyl-p-phenylenediamine, 97%
C10H16N2
306975
tert-Butyl methyl ether, anhydrous, 99.8%
C5H12O
675407
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
C5H12O
08603
tert-Butyl methyl ether, analytical standard
C5H12O
293210
tert-Butyl methyl ether, HPLC grade, for HPLC, 99.8%
C5H12O
PHR1560
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
C5H12O