EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Dohaekseunggi-tang extract inhibits obesity, hyperlipidemia, and hypertension in high-fat diet-induced obese mice.
PMID 25280587
Abstract
Dohaekseunggi-tang (DHSGT) is a traditional plant-based medicine prescribed to promote blood circulation and to treat obesity and hypertension. The present study aimed to identify potential anti-obesity activities of DHSGT extract. Anti-obesity, anti-hyperlipidemic, and anti-hypertensive effects of orally-administered DHSGT extract were evaluated in high-fat diet- (HFD)-induced obese mice. Serum biochemistry profiles and expression of diverse metabolic regulatory gene mRNAs in mouse visceral fat were assessed by RT-PCR. The effects of DHSGT on angiotensin-1 converting enzyme (ACE) and pancreatic lipase activities were determined using in vitro inhibition assays. Oral DHSGT treatment reduced obese HFD C57BL/6 J mouse body weight, liver and adipose tissue mass, adipocyte size, and blood pressure versus untreated HFD mice. DHSGT also decreased serum total cholesterol, LDL-cholesterol, triglyceride, glucose, and leptin concentrations, and increased HDL-cholesterol and adiponectin levels in HFD mice. Furthermore, DHSGT markedly increased mRNA expression of peroxisome proliferator activated receptor-gamma, uncoupling protein-2, and adiponectin in visceral adipose tissue of HFD mice. In vitro tests revealed that DHSGT effectively inhibited porcine pancreatic lipase and ACE activities, with IC50 values of 7.58 mg/ml and 0.56 mg/ml, respectively. These results validate traditional knowledge and suggest that DHSGT may be potentially useful for managing hyperlipidemia, hyperglycemia, hypertension, and obesity.