EMAIL THIS PAGE TO A FRIEND
International immunology
A novel function of interferon regulatory factor-1: inhibition of Th2 cells by down-regulating the Il4 gene during Listeria infection.
PMID 25280793
Abstract
Infection with certain pathogens induces a shift of the Th subset balance to a Th1 dominant state. This, in turn, results in the suppression of Th2 responses. We focused on the involvement of interferon regulatory factor-1 (IRF-1) in the suppression of Th2 cells during Listeria infection. We found that the inhibition of IL-4 production by Th2 cells is mediated by a soluble factor (LmSN) produced by Listeria-infected antigen-presenting cells. The inhibition is not observed with T cells from Irf1 gene-targeted mice. IRF-1 suppresses transcription of the Il4 gene in Th2 cells. Under the influence of the LmSN, IRF-1 binds to the 3' untranslated region (UTR) region of the Il4 gene and down-regulates Il4 gene transcription. Finally, we identified IL-1α and IL-1β as the mediator of the LmSN activity. Signaling through IL-1R induces the stabilization and/or nuclear translocation of IRF-1. We propose that IRF-1 functions to induce the T-cell subset shift via a novel mechanism. Under the influence of IL-1, IRF-1 translocates into the nucleus and acts on the 3'UTR region of the Il4 gene, thus inhibiting its transcription in Th2 cells. As a result, the immune system shifts predominantly to a Th1 response during Listeria infection, resulting in effective protection of the host.