EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Improvement of neuronal bioenergetics by neurosteroids: implications for age-related neurodegenerative disorders.
PMID 25281013
Abstract
The brain has high energy requirements to maintain neuronal activity. Consequently impaired mitochondrial function will lead to disease. Normal aging is associated with several alterations in neurosteroid production and secretion. Decreases in neurosteroid levels might contribute to brain aging and loss of important nervous functions, such as memory. Up to now, extensive studies only focused on estradiol as a promising neurosteroid compound that is able to ameliorate cellular bioenergetics, while the effects of other steroids on brain mitochondria are poorly understood or not investigated at all. Thus, we aimed to characterize the bioenergetic modulating profile of a panel of seven structurally diverse neurosteroids (progesterone, estradiol, estrone, testosterone, 3α-androstanediol, DHEA and allopregnanolone), known to be involved in brain function regulation. Of note, most of the steroids tested were able to improve bioenergetic activity in neuronal cells by increasing ATP levels, mitochondrial membrane potential and basal mitochondrial respiration. In parallel, they modulated redox homeostasis by increasing antioxidant activity, probably as a compensatory mechanism to a slight enhancement of ROS which might result from the rise in oxygen consumption. Thereby, neurosteroids appeared to act via their corresponding receptors and exhibited specific bioenergetic profiles. Taken together, our results indicate that the ability to boost mitochondria is not unique to estradiol, but seems to be a rather common mechanism of different steroids in the brain. Thus, neurosteroids may act upon neuronal bioenergetics in a delicate balance and an age-related steroid disturbance might be involved in mitochondrial dysfunction underlying neurodegenerative disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6546
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
C10H12O4
A7755
5α-Androstane-3α,17β-diol
C19H32O2
P8887
5α-Pregnan-3α-ol-20-one, solid
C21H34O2
E9750
Estrone, ≥99%
C18H22O2
E1274
Estrone, meets USP testing specifications
C18H22O2
46573
Estrone, VETRANAL, analytical standard
C18H22O2
E1700000
Estrone, European Pharmacopoeia (EP) Reference Standard
C18H22O2
1255001
Estrone, United States Pharmacopeia (USP) Reference Standard
C18H22O2
PHR1535
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material
C18H22O2
H4166
Hydroxyflutamide, ≥98% (HPLC)
C11H11F3N2O4
M8046
Mifepristone, ≥98%
C29H35NO2
1568007
Progesterone, United States Pharmacopeia (USP) Reference Standard
C21H30O2
46665
Progesterone, VETRANAL, analytical standard
C21H30O2
PHR1142
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
C21H30O2
P3300000
Progesterone, European Pharmacopoeia (EP) Reference Standard
C21H30O2
Y0001666
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H30O2
Y0001665
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
C21H30O2
T1500
Testosterone, ≥98%
C19H28O2
T0100000
Testosterone, European Pharmacopoeia (EP) Reference Standard
C19H28O2
46923
Testosterone, VETRANAL, analytical standard
C19H28O2
86500
Testosterone, purum, ≥99.0% (HPLC)
C19H28O2
Y0000342
Testosterone for impurity D identification, European Pharmacopoeia (EP) Reference Standard
C19H28O2
Y0000343
Testosterone for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H28O2
T-037
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C19H28O2